Skip to Content
Merck
All Photos(2)

Documents

244422

Sigma-Aldrich

1-Hexyne

97%

Synonym(s):

Butylacetylene, n-Butylacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3C≡CH
CAS Number:
Molecular Weight:
82.14
Beilstein:
635687
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39010407
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

253 mmHg ( 37.7 °C)

Assay

97%

form

liquid

impurities

<2% 1-bromobutane

refractive index

n20/D 1.399 (lit.)

bp

71-72 °C (lit.)

mp

−132 °C (lit.)

density

0.715 g/mL at 25 °C (lit.)

SMILES string

CCCCC#C

InChI

1S/C6H10/c1-3-5-6-4-2/h1H,4-6H2,2H3

InChI key

CGHIBGNXEGJPQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hexyne reacted with thianthrene cation radical tetrafluoroborate to form trans-1,2-bis(5-thianthreniumyl)alkene tetrafluoroborate.

Application

1-Hexyne was used in the synthesis of a tricyclic isoindolinone scaffold by undergoing hydrozirconation and ring-closing metathesis (RCM).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bettina Miller et al.
Beilstein journal of organic chemistry, 8, 1091-1097 (2012-09-29)
Hydrozirconation of 1-hexyne, the addition to in situ prepared N-acyliminium species, and ring-closing metathesis (RCM) were key steps in the preparation of a tricyclic isoindolinone scaffold. An unusual alkene isomerization process during the RCM was identified and studied in some
David A Bichara et al.
The Journal of arthroplasty, 33(8), 2666-2670 (2018-05-23)
Wear resistance of ultrahigh molecular weight polyethylene (UHMWPE) is improved via ionizing radiation crosslinking and subsequent high temperature melting for improved toughness. Our group has previously reported that crosslinking can also be achieved chemically using organic peroxides. However, volatile peroxide
Mauricio M Victor et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 15(4), 400-408 (2018-10-27)
The trypanosomatids, such as the protozoan Leishmania spp., have a demand by ergosterol, which is not present in the membrane from mammal cells. The suppression of the synthesis of ergosterol would be a new target of compounds with leishmanicidal activity
Hyomin Jin et al.
Molecules (Basel, Switzerland), 24(1) (2019-01-10)
Herein, we investigated the effect of ring planarity by fully characterizing four pyridine-based o-carboranyl compounds. o-Carborane was introduced to the C4 position of the pyridine rings of 2-phenylpyridine and 2-(benzo[b]thiophen-2-yl)pyridine (CB1 and CB2, respectively), and the compounds were subsequently borylated
He Nan et al.
Journal of chromatography. A, 1523, 316-320 (2017-06-26)
Silver ion or argentation chromatography utilizes stationary phases containing silver ions for the separation of unsaturated compounds. In this study, a mixed-ligand silver-based ionic liquid (IL) was evaluated for the first time as a gas chromatographic (GC) stationary phase for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service