Skip to Content
Merck
All Photos(1)

Documents

253197

Sigma-Aldrich

Dibenzo-24-crown-8

98%

Synonym(s):

2,3,14,15-Dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosa-2,14-diene, 6,7,9,10,12,13,20,21,23,24,26,27-Dodecahydrodibenz[b,n]-1,4,7,10,13,16,19,22-octaoxacyclotetracosane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32O8
CAS Number:
Molecular Weight:
448.51
Beilstein:
1693744
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

103-105 °C (lit.)

SMILES string

C1COCCOc2ccccc2OCCOCCOCCOc3ccccc3OCCO1

InChI

1S/C24H32O8/c1-2-6-22-21(5-1)29-17-13-25-9-10-27-15-19-31-23-7-3-4-8-24(23)32-20-16-28-12-11-26-14-18-30-22/h1-8H,9-20H2

InChI key

UNTITLLXXOKDTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibenzo-24-crown-8 is a macrocyclic compound that is used as a versatile reagent in organic synthesis due to its ability to form complexes with metal ions and enhance their solubility and reactivity. It is also used as a phase-transfer catalyst in various organic reactions, such as the synthesis of organic carbonates and the alkylation of phenols.

Application

Dibenzo-24-crown-8 can be used as a molecular transporter in the formation of [1]rotaxanes with thiazolium-based mono- and tris-branched ammonium ions. It is also used as a co-solvent and phase-transfer catalyst for cesium fluoride, to the development of a more efficient anhydrous fluorinating system for organic synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mateusz Woźny et al.
Dalton transactions (Cambridge, England : 2003), 48(19), 6546-6557 (2019-04-24)
[2]rotaxanes composed of a dibenzo-24-crown-8 wheel (DB24C8) and an axle containing two metal-complexing tetraazamacrocyclic units linked with a p-xylylene bridge, coordinating the same or different metal ions (Ni and/or Cu) are described. A symmetric di-Ni rotaxane crystallizes in the monoclinic
Nozar Moradi et al.
Analytica chimica acta, 1108, 118-128 (2020-03-31)
Most attributes of the bulk materials, especially in the solid-state, are directly dictated by a manner by which the molecules are ordered. Thus, it is expected that the possibility of controlling these structural orders would allow predominating some particular physical
Lei Fang et al.
Journal of the American Chemical Society, 131(20), 7126-7134 (2009-05-08)
A versatile synthetic strategy, which was conceived and employed to prepare doubly threaded, bistable [c2]daisy chain compounds, is described. Propargyl and 1-pentenyl groups have been grafted onto the stoppers of [c2]daisy chain molecules obtained using a template-directed synthetic protocol. Such
Frédéric Coutrot et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(21), 5186-5190 (2009-02-21)
Molecular machines boarding now! Molecular machines containing dibenzo[24]crown-8 (DB24C8) and based on an anilinium and a pyridinium amide station have been prepared. The DB24C8 shuttles upon variation of pH and interacts differently with the pyridinium amide station depending on its
Yu-Ying Huang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 243, 118754-118754 (2020-08-20)
As a colorimetric and fluorescent turn-on sensor to Al3+, N'-(2-hydroxybenzylidene)isoquinoline-3-carbohydrazide (HL) has been easily synthesized. The fluorescence intensity increases by 273 times in the presence of Al3+ at 458 nm. Meanwhile, the experiment data indicate that the limit of detection for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service