Skip to Content
Merck
All Photos(1)

Documents

251569

Sigma-Aldrich

1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone

98%, for peptide synthesis

Synonym(s):

1,3-Dimethyl-2-oxohexahydropyrimidine, N,N′-Dimethylpropylene urea, DMPU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O
CAS Number:
Molecular Weight:
128.17
Beilstein:
110562
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

product name

1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 98%

Assay

98%

form

liquid

refractive index

n20/D 1.488 (lit.)

bp

146 °C/44 mmHg (lit.)

density

1.06 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

CN1CCCN(C)C1=O

InChI

1S/C6H12N2O/c1-7-4-3-5-8(2)6(7)9/h3-5H2,1-2H3

InChI key

GUVUOGQBMYCBQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone (DMPU) can be used:
  • As a versatile solvent in N-alkylation of amines
  • O-alkylation of aldoses , and in the synthesis of poly(aryl ethers).
  • As urea solvent in the synthesis of 1-aryl-3,4,5-substituted pyrazoles by cyclocondensation of arylhydrazine hydrochlorides with 1,3-diketones.
  • As an additive in asymmetric cyanoamidation.

DMPU/HF can be used as a nucleophilic fluorinating agent in fluoro-Prins reaction for the synthesis of substituted 4-fluorotetrahydropyrans and 4-fluoropiperidines.
Versatile solvent employed in N-alkylation of chiral, primary amines, O-alkylation of aldoses, and in the synthesis of poly(aryl ethers).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thianthrene as an activating group for the synthesis of poly (aryl ether thianthrene)s by nucleophilic aromatic substitution
Edson JB and Knauss DM
Journal of Polymer Science Part A: Polymer Chemistry, 42(24), 6353-6363 (2004)
Intermolecular N-alkylation of amines under conditions of the Mitsunobu reaction: a new solid-phase synthesis of tertiary benzylamines
Zaragoza, Florencio and Stephensen, Henrik
Tetrahedron Letters, 41(11), 1841-1844 (2000)
Protecting Groups: Strategies and Applications in Carbohydrate Chemistry, 41(11), 337-370 (2019)
Synthesis, 1243-1243 (1993)
Highly Enantioselective Intramolecular Cyanoamidation:(+)-Horsfiline,(-)-Coerulescine, and (-)-Esermethole
Reddy VJ and Douglas CJ
Organic Letters, 12(5), 952-955 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service