Skip to Content
Merck
All Photos(3)

Key Documents

248762

Sigma-Aldrich

Silver nitrate on silica gel

extent of labeling: ~10 wt. % loading, +230 mesh

Synonym(s):

Nitric acid silver(I) salt, Silver (I) nitrate, Silver mononitrate, Silver nitrate (AgNO3)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

core: silver
reagent type: catalyst

extent of labeling

~10 wt. % loading

particle size

+230 mesh

solubility

water: soluble 216g/100g at 25 °C(lit.)
benzene: soluble 2.2 g/L(lit.)
ethanol: soluble 20.8 g/L(lit.)
methanol: soluble 35 g/L(lit.)

density

4.35 g/mL at 25 °C (lit.)

functional group

nitrate

SMILES string

[Ag+].[O-][N+]([O-])=O

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Silver nitrate on silica gel has been used in determination of alkene-based drilling fluids in crude oils by silica gel and silver-impregnated silica gel chromatography.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher M Reddy et al.
Journal of chromatography. A, 1148(1), 100-107 (2007-03-23)
Comprehensive two-dimensional gas chromatography with flame ionization detection (GC x GC-FID) was used to measure alkene-based drilling fluids in crude oils. Compared to one-dimensional gas chromatography, GC x GC-FID is more robust for detecting alkenes due to the increased resolution
Ali Hebeish et al.
Carbohydrate polymers, 92(1), 407-413 (2012-12-12)
Innovative CMC-based hydrogels with great potentials for usage in medical area were principally synthesized as per two strategies .The first involved reaction of epichlorohydrin in alkaline medium containing silver nitrate to yield silver nano-particles (AgNPs)-loaded CMC hydrogel. While CMC acted
Tao Xu et al.
Organic letters, 14(21), 5416-5419 (2012-10-24)
A silver-catalyzed intramolecular oxidative aminofluorination of alkynes has been developed by using NFSI as a fluorinating reagent. This reaction represents an efficient method for the synthesis of various 4-fluoroisoquinolines and 4-fluoropyrrolo[α]isoquinolines.
Benjamin P Colman et al.
PloS one, 8(2), e57189-e57189 (2013-03-08)
A large fraction of engineered nanomaterials in consumer and commercial products will reach natural ecosystems. To date, research on the biological impacts of environmental nanomaterial exposures has largely focused on high-concentration exposures in mechanistic lab studies with single strains of
Irina Blinova et al.
Environmental science and pollution research international, 20(5), 3456-3463 (2012-11-13)
Although silver nanoparticles (NPs) are increasingly used in various consumer products and produced in industrial scale, information on harmful effects of nanosilver to environmentally relevant organisms is still scarce. This paper studies the adverse effects of silver NPs to two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service