Skip to Content
Merck
All Photos(3)

Key Documents

248622

Sigma-Aldrich

Silicon nitride

predominantly β-phase, ≤10micron primary particle size

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si3N4
CAS Number:
Molecular Weight:
140.28
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder

primary particle size

≤10micron

density

3.44 g/mL at 25 °C (lit.)

SMILES string

N12[Si]34N5[Si]16N3[Si]25N46

InChI

1S/N4Si3/c1-5-2-6(1)3(5)7(1,2)4(5)6

InChI key

HQVNEWCFYHHQES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Christophe Baker et al.
Optics express, 20(27), 29076-29089 (2012-12-25)
We report time domain observations of optical instability in high Q silicon nitride whispering gallery disk resonators. At low laser power the transmitted optical power through the disk looks chaotic. At higher power, the optical output settles into a stable
Benjamin N Miles et al.
Chemical Society reviews, 42(1), 15-28 (2012-09-20)
This tutorial review will introduce and explore the fundamental aspects of nanopore (bio)sensing, fabrication, modification, and the emerging technologies and applications that both intrigue and inspire those working in and around the field. Although nanopores can be classified into two
Kapil Debnath et al.
Optics express, 20(25), 27420-27428 (2012-12-25)
Integration density, channel scalability, low switching energy and low insertion loss are the major prerequisites for on-chip WDM systems. A number of device geometries have already been demonstrated that fulfill these criteria, at least in part, but combining all of
Orsolya Tapasztó et al.
Journal of nanoscience and nanotechnology, 12(11), 8775-8778 (2013-02-21)
The dispersion properties of single- and multi-walled carbon nanotubes as well as mechanically exfoliated few layer graphene flakes within the silicon nitride ceramic matrix have been investigated. Small angle neutron scattering experiments have been employed to gain information on the
Feng Wang et al.
Optics express, 21(2), 1675-1686 (2013-02-08)
A maximal enhancement of ~6.5 times of the external quantum efficiency (EQE) for SiNx-based light-emitting devices (LEDs) is achieved by magnetron sputtering a silver nanostructures layer onto the active matrix. The enhancement of EQE is affected by the dimension and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service