All Photos(1)
About This Item
Linear Formula:
BrCH2CH2COOCH3
CAS Number:
Molecular Weight:
167.00
Beilstein:
1742949
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.458 (lit.)
bp
64-66 °C/18 mmHg (lit.)
density
1.53 g/mL at 25 °C (lit.)
functional group
bromo
ester
SMILES string
COC(=O)CCBr
InChI
1S/C4H7BrO2/c1-7-4(6)2-3-5/h2-3H2,1H3
InChI key
KQEVIFKPZOGBMZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Methyl 3-bromopropionate was used in the synthesis of spiroanellated γ-lactones, pinacols and unprecedented 3-(1-hydroxycycloalkyl)-l-oxaspiro[n,m]alkan-2-ones.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
167.0 °F - closed cup
Flash Point(C)
75 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Samarium diiodide promoted spirolactonization of cycloalkanones.
Hua Z, et al.
Tetrahedron, 47(34), 7037-7044 (1991)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service