All Photos(2)
About This Item
Linear Formula:
CH3CH2CHBrCOOC2H5
CAS Number:
Molecular Weight:
195.05
Beilstein:
969841
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.447 (lit.)
bp
177 °C (lit.)
density
1.321 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CCOC(=O)C(Br)CC
InChI
1S/C6H11BrO2/c1-3-5(7)6(8)9-4-2/h5H,3-4H2,1-2H3
InChI key
XIMFCGSNSKXPBO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
132.8 °F - closed cup
Flash Point(C)
56 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Daria Kupczyk et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-19)
Glucocorticoid metabolism at the tissue level is regulated by two isoenzymes 11β-hydroxysteroid dehydrogenase (11β-HSD), which mutually convert biologically active cortisol and inactive cortisone. Recent research is focused on the role of 11β-HSD1 and 11β-HSD2 as autocrine factors of tumor cell
Renata Studzińska et al.
Chemistry & biodiversity, 16(6), e1900065-e1900065 (2019-04-24)
11β-Hydroxysteroid dehydrogenase type 1 (11β-HSD1) is an enzyme that affects the body's cortisol levels. The inhibition of its activity can be used in the treatment of Cushing's syndrome, metabolic syndrome and type 2 diabetes. In this study, we synthesized new
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service