Skip to Content
Merck
All Photos(2)

Key Documents

236241

Sigma-Aldrich

Pentaerythritol

99%

Synonym(s):

2,2-Bis(hydroxymethyl)-1,3-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C(CH2OH)4
CAS Number:
Molecular Weight:
136.15
Beilstein:
1679274
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<1 mmHg ( 20 °C)

Assay

99%

bp

276 °C/30 mmHg (lit.)

mp

253-258 °C (lit.)

SMILES string

OCC(CO)(CO)CO

InChI

1S/C5H12O4/c6-1-5(2-7,3-8)4-9/h6-9H,1-4H2

InChI key

WXZMFSXDPGVJKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentaerythritol is acrystalline , odour less white solid. The presence of neopentane core and four terminalhydroxyl groups makes it a versatile substrate for the synthesispolyfunctionalized compounds.

Application

Pentaerythritol can be used as a starting material to prepare four armed polyurethane prepolymers, which are applicable in the synthesis of UV cured polyurethane dispersions.

It can be used as an additive in the preparation of flame retardant polymers.

It can also be used as a precursor to synthesize polyethylene glycol/4,4′-diphenylmethane diisocyanate/pentaerythritol copolymer which is applicable as heat storage material.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chin-Wen Chen et al.
Polymers, 12(9) (2020-09-10)
Bio-based unsaturated poly(butylene adipate-co-butylene itaconate) (PBABI) aliphatic copolyesters were synthesized with pentaerythritol (PE) as a modifier, observing the melting point, crystallization, and glass transition temperatures were decreased from 59.5 to 19.5 °C and 28.2 to -9.1 °C as an increase
Mohamed Touaibia et al.
Chemical communications (Cambridge, England), (4)(4), 380-382 (2007-01-16)
Several oligomannoside clusters having a hundred-fold increase in affinities toward E. coli were synthesized by Cu(I)-catalyzed [1,3]-dipolar cycloadditions using pentaerythritol scaffolds bearing either alkyne or azide functionalities.
Samar Hamdy et al.
Journal of pharmaceutical and biomedical analysis, 44(4), 914-923 (2007-06-26)
The present study had two main objectives. First, was to compare the immune stimulatory effect of two synthetic lipid A analogues (7-acyl lipid A and pentaerythritol-based lipid A (PET lipid A)) on maturation/stimulation of bone marrow derived dendritic cells (DCs).
Michael C Hacker et al.
Biomacromolecules, 9(6), 1558-1570 (2008-05-17)
In this study, we synthesized and characterized a series of macromers based on poly( N-isopropylacrylamide) that undergo thermally induced physical gelation and, following chemical modification, can be chemically cross-linked. Macromers with number average molecular weights typically ranging from 2000-3500 Da
Tao Tu et al.
The Journal of organic chemistry, 73(14), 5255-5263 (2008-06-19)
Molecular scaffolds that have well-defined geometries, are easy to synthesize and functionalize, and can hold attached sites of molecular recognition in suitable orientations are useful tools in various areas of science and technology. The utility of the tetraphenyl ether of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service