Skip to Content
Merck
All Photos(3)

Key Documents

235865

Sigma-Aldrich

Butyl methacrylate

99%, contains monomethyl ether hydroquinone as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COO(CH2)3CH3
CAS Number:
Molecular Weight:
142.20
Beilstein:
773960
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

4.91 (15 °C, vs air)

vapor pressure

2 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

562 °F

contains

monomethyl ether hydroquinone as inhibitor

expl. lim.

2-8 %

refractive index

n20/D 1.423 (lit.)

bp

162-165 °C (lit.)

density

0.894 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCOC(=O)C(C)=C

InChI

1S/C8H14O2/c1-4-5-6-10-8(9)7(2)3/h2,4-6H2,1,3H3

InChI key

SOGAXMICEFXMKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl methacrylate is an organic compound that belongs to the class of methacrylate esters and is commonly used in the production of various polymer materials. It can be copolymerized with other monomers to synthesize copolymers with tailored properties. The resulting copolymers find use in numerous fields, including automotive coatings, textiles, adhesives, contact lenses, and dental resins.

Application

Butyl methacrylate (BMA) can be used as:
  • A Monomer to synthesize waterborne butyl methacrylate (co)polymers latexes via emulsion polymerization for the development of coating materials for slow release-fertilizers.
  • A matrix polymer in the preparation of LDH/poly (butyl methacrylate) nanocomposites, which can improve the mechanical properties and thermal stability of the resulting nanocomposites.
  • A monomer to synthesize styrene and butyl methacrylate copolymers for use in leather finishing. Increasing the BMA content of the copolymer increases the water resistance and decreases the water vapor permeability of the leather.
  • A monomer for the preparation of monolithic columns on microfluidic chips. These columns can be used for a wide range of applications, such as the separation and analysis of complex mixtures of compounds, the preconcentration of samples, and the development of new microfluidic analytical devices.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

119.3 °F - closed cup

Flash Point(C)

48.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R-G Li et al.
European review for medical and pharmacological sciences, 19(12), 2213-2220 (2015-07-15)
The recent PLATINUM trial has demonstrated that the use of the new generation platinum chromium everolimus-eluting stents (PtCr-EES) yield clinical outcomes similar to those obtained by the use of cobalt chromium everolimus-eluting stents (CoCr-EES) in selected patients with 1 or
K Ishihara et al.
Journal of biomaterials applications, 13(2), 111-127 (1998-10-20)
We have synthesized phospholipid polymers containing 2-methacryloyloxyethyl phosphorylcholine (MPC) units as novel blood compatible polymers and have evaluated their interactions with blood components. It was found that in the absence of anticoagulants, blood clotting was delayed and blood cell adhesion
R Yoshida et al.
Journal of biomaterials science. Polymer edition, 3(2), 155-162 (1991-01-01)
Thermosensitive co-polymers of isopropyl acrylamide (IPAAm) with butyl methacrylate (BMA) are capable of 'on-off' regulation of drug release in response to external temperature changes due to skin formation with increasing temperature. To clarify the role of the surface-modulated skin and
Ke Li et al.
Electrophoresis, 40(16-17), 2142-2148 (2019-04-30)
In this study, an in-tube solid-phase microextraction column packed with mesoporous TiO2 nanoparticles, coupled with MALDI-TOF-MS, was applied to the selective enrichment and detection of phosphopeptides in complex biological samples. The mesoporous TiO2 nanoparticles with high specific surface areas, prepared
Mitsuyoshi Yamane et al.
Macromolecular rapid communications, 41(18), e2000141-e2000141 (2021-01-20)
Poly(n-butyl methacrylate) (PnBMA)/reduced graphene oxide (rGO) nanocomposite films are prepared using two different routes. The first route involves preparation of PnBMA nanoparticles containing homogeneously dispersed rGO nanosheets by miniemulsion polymerization using a block copolymer of ionic liquid (IL) monomer and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service