Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Butyl methacrylate
Synonym(s):
Butyl methacrylate
Linear Formula:
CH
2
=C(CH
3
)COO(CH
2
)
3
CH
3
CAS Number:
97-88-1
Molecular Weight:
142.20
EC Number:
202-615-1
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (2)
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Monomer Type
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
235865
Butyl methacrylate,
99%, contains monomethyl ether hydroquinone as inhibitor
Expand
View Pricing
8.00585
Butyl methacrylate,
(stabilised) for synthesis
Expand
View Pricing
Y0001487
Butyl methacrylate,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing