Skip to Content
Merck
All Photos(1)

Key Documents

234478

Sigma-Aldrich

1-Chloro-3-iodopropane

99%

Synonym(s):

Trimethylene chloroiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)3I
CAS Number:
Molecular Weight:
204.44
Beilstein:
1731115
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
99%

vapor density

6.8 (vs air)

Quality Level

vapor pressure

2 mmHg ( 20 °C)

Assay

99%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.548 (lit.)

bp

170-172 °C (lit.)

density

1.904 g/mL at 25 °C (lit.)

functional group

chloro
iodo

SMILES string

ClCCCI

InChI

1S/C3H6ClI/c4-2-1-3-5/h1-3H2

InChI key

SFOYQZYQTQDRIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Chloro-3-iodopropane undergoes asymmetric α-alkylation with N-sulfinyl imidates to yield 2-substituted N-tert-butanesulfinyl-5-chloropentanimidates. Electroreduction of 1-chloro-3-iodopropane at glassy carbon electrode in dimethylformamide containing tetra-n-butylammonium perchlorate has been investigated by cyclic voltammetry. It also participates in conjugate addition of alkyl iodides to α,β-unsaturated nitriles in water.

Application

1-Chloro-3-iodopropane has been used in the synthesis of:
  • N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid, an inhibitor of glycinamide ribonucleotide transformylase (GAR Tfase) and aminoimidazole carboxamide ribonucleotide transformylase (AICAR Tfase)
  • interesting ″proton sponge″ type molecule quino[7,8-h]quinoline

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical Reduction of 1, 3-Dihalopropanes at Carbon Cathodes in Dimethylformamide.
Pritts WA and Peters DG.
Journal of the Electrochemical Society, 141(4), 990-995 (1994)
Fraser F Fleming et al.
The Journal of organic chemistry, 72(18), 6961-6969 (2007-08-10)
A new silica-supported zinc-copper matrix reagent promotes the conjugate addition of alkyl iodides to cyclic and acyclic alkenenitriles in water. X-ray diffraction and electron microscopy techniques suggest that the active copper species generated from elemental zinc and copper(I) iodide is
Synthesis of 1, 2, 3, 4-tetrahydroquinolines and 1, 2, 3, 4-tetrahydro-1, 6-naphthyridines by a directed lithiation reaction.
Reed JN, et al.
Tetrahedron Letters, 29(45), 5725-5728 (1988)
Filip Colpaert et al.
The Journal of organic chemistry, 76(1), 234-244 (2010-12-02)
α-Alkylation of N-sulfinyl imidates with 1-chloro-3-iodopropane successfully led to 2-substituted N-tert-butanesulfinyl-5-chloropentanimidates in acceptable diastereomeric ratios (dr 67/33 to 72/28) and good yields (74-86%). Subsequent reduction with NaBH(4) led to the corresponding 2-substituted N-tert-butanesulfinyl-5-chloropentylamines, which could be cyclized to a range
Heng Cheng et al.
Bioorganic & medicinal chemistry, 13(10), 3593-3599 (2005-04-26)
The synthesis and evaluation of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid (2) as an inhibitor of glycinamide ribonucleotide transformylase (GAR Tfase) and aminoimidazole carboxamide ribonucleotide transformylase (AICAR Tfase) are reported. The inhibitor 2 was prepared in a convergent synthesis involving C-alkylation of methyl 4-(4,4,4-trifluoro-3-dimethylhydrazonobutyl)benzoate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service