232467
1,11-Dibromoundecane
≥98%
Synonym(s):
Undecamethylene dibromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Br(CH2)11Br
CAS Number:
Molecular Weight:
314.10
Beilstein:
1740980
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥98%
form
liquid
refractive index
n20/D 1.491 (lit.)
bp
190-192 °C/18 mmHg (lit.)
mp
−10.6 °C (lit.)
density
1.335 g/mL at 25 °C (lit.)
functional group
bromo
SMILES string
BrCCCCCCCCCCCBr
InChI
1S/C11H22Br2/c12-10-8-6-4-2-1-3-5-7-9-11-13/h1-11H2
InChI key
SIBVHGAPHVRHMJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
1,11-Dibromoundecane was used in preparation of:
- thermotropic main-chain polyether and has been studied by variable-temperature solid-state 13C NMR spectroscopy
- discotic side group liquid crystal polymer based on poly(methyl methacrylate)
- self-assembled monolayer (SAM) substrate, 11-(mercaptoundecyl)triethylene glycol
- SAM of a ferrocene (Fc)-porphyrin (P)-C60 triad on a gold electrode
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Convenient synthesis of a discotic side group liquid crystal polymer.
Mchattie GS and Imrie CT.
Journal of Materials Chemistry, 8(1), 47-51 (1998)
The chain conformations of a polyether based on bis (4-hydroxyphenoxy)-p-xylylene and 1, 11-dibromoundecane (HPX-c11) in its mesophase states, derived from 13C NMR.
Cheng J, et al.
Journal of Polymer Science. Part B, Polymer Letters, 32(4), 721-736 (1994)
New syntheses for 11-(mercaptoundecyl) triethylene glycol and mercaptododecyltriethyleneoxy biotin amide.
Canaria CA, et al.
Tetrahedron Letters, 46(28), 4813-4816 (2005)
Synthesis and photoelectrochemical properties of a self-assembled monolayer of a ferrocene-porphyrin-fullerene triad on a gold electrode.
Imahori H, et al.
Chemical Communications (Cambridge, England), 13, 1165-1166 (1999)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service