Skip to Content
Merck
All Photos(1)

Documents

144991

Sigma-Aldrich

1,7-Dibromoheptane

97%

Synonym(s):

Heptamethylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)7Br
CAS Number:
Molecular Weight:
257.99
Beilstein:
1734575
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.503 (lit.)

bp

255 °C (lit.)

density

1.51 g/mL at 25 °C (lit.)

SMILES string

BrCCCCCCCBr

InChI

1S/C7H14Br2/c8-6-4-2-1-3-5-7-9/h1-7H2

InChI key

LVWSZGCVEZRFBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,7-Dibromoheptane was used in the synthesis of random thermotropic liquid crystalline copolyether and ternary copolyether. It was also used in the synthesis of thermotropic liquid crystalline dendrimer exhibiting a nematic phase.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid crystalline polyethers based on conformational isomerism. 6. Influence of copolymer composition of a ternary copolyether based on 1-(4-hydroxyphenyl)-2-(2-methyl-4-hydroxyphenyl) ethane, 1, 5-dibromopentane, 1, 7-dibromoheptane, and 1, 9-dibromononane on its mesomorphic phase transitions.
Percec V and Tsuda Y.
Macromolecules, 23(1), 5-12 (1990)
Synthesis and characterization of a thermotropic nematic liquid crystalline dendrimeric polymer.
Percec V and Kawasumi M.
Macromolecules, 25(15), 3843-3850 (1992)
Yuan-Ping Pang et al.
PloS one, 4(2), e4349-e4349 (2009-02-06)
Aphids, among the most destructive insects to world agriculture, are mainly controlled by organophosphate insecticides that disable the catalytic serine residue of acetylcholinesterase (AChE). Because these agents also affect vertebrate AChEs, they are toxic to non-target species including humans and
Functional polymers and sequential copolymers by phase transfer catalysis. 24. The influence of molecular weight on the thermotropic properties of a random copolyether based on 1, 5-dibromopentane, 1, 7-dibromoheptane, and 4, 4'-dihydroxy-a-methylstilbene.
Percec V, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 25(7), 1943-1965 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service