Skip to Content
Merck
All Photos(1)

Documents

23000

Sigma-Aldrich

2-Chloroethanol

puriss. p.a., ≥99.0% (GC)

Synonym(s):

Ethylene chlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2OH
CAS Number:
Molecular Weight:
80.51
Beilstein:
878139
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.78 (vs air)

vapor pressure

5 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

797 °F

expl. lim.

16 %

impurities

≤0.5% water

refractive index

n20/D 1.441 (lit.)
n20/D 1.442

bp

129 °C (lit.)

mp

−89 °C (lit.)

density

1.201 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤10.0 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.1 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.5 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤2 mg/kg
Ni: ≤0.1 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

OCCCl

InChI

1S/C2H5ClO/c3-1-2-4/h4H,1-2H2

InChI key

SZIFAVKTNFCBPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hanah Mesallati et al.
Pharmaceutics, 11(6) (2019-06-12)
Enrofloxacin (ENRO) is a poorly soluble drug used in veterinary medicine. It differs from the more widely used fluoroquinolone ciprofloxacin (CIP) by the presence of an ethyl substituent on its piperazine amino group. While a number of recent studies have
Rose Roberts et al.
Materials science & engineering. C, Materials for biological applications, 108, 110191-110191 (2020-01-12)
Effective therapeutic delivery of peptide and protein drugs is challenged by short in vivo half-lives due to rapid degradation. Sustained release formulations of αCT1, a 25 amino acid peptide drug, would afford lower dosing frequency in indications that require long
Apryl Bronley-DeLancey et al.
Environmental health perspectives, 114(8), 1237-1242 (2006-08-03)
Trichloroethylene (TCE) is a suspected human carcinogen and a common groundwater contaminant. Chloral hydrate (CH) is the major metabolite of TCE formed in the liver by cytochrome P450 2E1. CH is metabolized to the hepatocarcinogen trichloroacetate (TCA) by aldehyde dehydrogenase
Mariella Carrieri et al.
Archives of toxicology, 81(7), 529-532 (2007-02-08)
Nonfatal acute inhalation of trichloroethylene (TRI) at work was described. The subject, male, 54 years old, was drawn unconscious by a metal-degreasing machine and immediately sheltered in intensive care unit. Other than basic life support and common laboratory indices, blood
Ronan Garlantézec et al.
Occupational and environmental medicine, 69(1), 62-70 (2011-06-28)
To describe urine levels of metabolites of glycol ethers and chlorinated solvents in a sample of pregnant women from the general population, to study their occupational and non-occupational determinants and to compare them with the results of indirect assessment methods

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service