Skip to Content
Merck
All Photos(2)

Key Documents

229555

Sigma-Aldrich

Cerium(III) fluoride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Cerous fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CeF3
CAS Number:
Molecular Weight:
197.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
powder

grade

anhydrous

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: cerium

impurities

≤150.0 ppm Trace Rare Earth Analysis

density

6.16 g/mL at 25 °C (lit.)

SMILES string

F[Ce](F)F

InChI

1S/Ce.3FH/h;3*1H/q+3;;;/p-3

InChI key

QCCDYNYSHILRDG-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hui Zhang et al.
Journal of colloid and interface science, 302(2), 509-515 (2006-08-01)
In this article, we firstly reported on the synthesis and characterization of ultrafine CeF3 nanoparticles (NPs) modified by catanionic surfactant via a reverse micelles-based route. The catanionic surfactant PN was prepared by mixing the di(2-ethylhexyl) phosphoric acid (DEHPA) and primary
G Wu et al.
Guang pu xue yu guang pu fen xi = Guang pu, 20(4), 560-562 (2003-08-30)
The matrix K3AlF6 was prepared with a reaction of mixture of solution K2CO3 upon certain solution of Al(OH)3 plus HF (40%), dehydrating at 100 degrees C and annealing for 48 h at 600 degrees C. A series of phosphors for
Florian J Wegehaupt et al.
Archives of oral biology, 56(12), 1541-1547 (2011-07-09)
Ninety-six dentine samples were prepared from human premolars and randomly assigned to eight groups (G1-G8). Samples were treated for 30s with the following solutions: placebo (G1/G2), amine fluoride (Elmex fluid; G3/G4), cerium chloride (G5/G6) and combined fluoride/cerium chloride application (G7/G8).
A E Ivanov et al.
Meditsinskaia radiologiia, 34(1), 60-65 (1989-01-01)
Macro- and microtopography of the distribution of aspirated colloidal 144CeF3 in autographs of rabbit lung total sections were examined with the help of a monitoring-measuring device "Videoplan" and microcomputer. Quantitative assessment of photoemulsion-blackening areas at the site of radionuclide concentration
Xiongxin Dai et al.
Health physics, 101(2), 144-147 (2011-06-29)
A rapid bioassay method has been developed for the sequential measurements of actinides in human urine samples. The method involves actinide separation from a urine matrix by co-precipitation with hydrous titanium oxide (HTiO), followed by anion exchange and extraction chromatography

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service