Skip to Content
Merck
All Photos(1)

Key Documents

229539

Sigma-Aldrich

Calcium oxide

99.995% trace metals basis

Synonym(s):

Lime, Quicklime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaO
CAS Number:
Molecular Weight:
56.08
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99.995% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: calcium

loss

10% loss on ignition, 1000°C (absorbed H2O and CO2)

bp

2850 °C (lit.)

density

3.3 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Ca]

InChI

1S/Ca.O

InChI key

ODINCKMPIJJUCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Units are 10% in excess of stated weight to compensate for the loss of H2O and CO2

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vasilije Manovic et al.
Environmental science & technology, 46(22), 12720-12725 (2012-10-24)
This paper presents a novel method for reactivation of spent CaO-based sorbents from calcium looping (CaL) cycles for CO(2) capture. A spent Cadomin limestone-derived sorbent sample from a pilot-scale fluidized bed (FBC) CaL reactor is used for reactivation. The calcined
Mohammad Mahdi Najafpour et al.
Dalton transactions (Cambridge, England : 2003), 42(14), 5085-5091 (2013-02-12)
Inspired by Nature's catalyst, a nano-size layered manganese-calcium oxide showed a low overvoltage for water oxidation in acidic solutions, which is comparable to platinum.
Ebrahim Negaresh et al.
Chemosphere, 92(11), 1513-1519 (2013-05-15)
This paper investigates the effect of using recycled fiber (RCF) in newsprint production on the effluent quality and its treatability using membrane operations for internal and external recycling and reuse. Increased chemical usage in RCF for deinking had significant impact
Mahsa Naghshineh et al.
Food chemistry, 136(2), 472-478 (2012-11-06)
The application of high hydrostatic pressure technology for enzymatic extraction of pectin was evaluated. Cellulase and xylanase under five different combinations (cellulase/xylanase: 50/0, 50/25, 50/50, 25/50, and 0/50 U/g lime peel) at ambient pressure, 100 and 200 MPa were used
Arup Roy et al.
Journal of biomedical nanotechnology, 9(9), 1570-1578 (2013-08-29)
The high degree of microbial diseases and their multidrug resistant properties make the researchers to develop new class of antimicrobial agents. A modern and innovative approach of drug development is the use of metallic nanoparticles as new formulations of antimicrobial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service