Skip to Content
Merck
All Photos(3)

Key Documents

224227

Sigma-Aldrich

Titanium(IV) oxide, rutile

powder, <5 μm, ≥99.9% trace metals basis

Synonym(s):

Titanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: titanium

particle size

<5 μm

density

4.17 g/mL at 25 °C (lit.)

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Titanium(IV) oxide is a semiconductorphotocatalyst with a band gap of 3.0 eV. It is widely used in the field of photovoltaics,sensors, and environmental remediation due to its unique properties such ashigh reactivity, stability against photocorrosion, and ease of furtherfunctionalization.

Application

Titanium(IV) oxide (TiO2) and functionalized TiO2 can be used as a photocatalyst for overall water-splitting reactions and photocatalytic hydrogen evolution.

Rutile titanium(IV) oxide exhibit a high dielectric constant and can be used as a component in low-temperature cofired ceramics (LTCC) or as a filler in organic-inorganic composites.

Au/rutileTiO2 can be used as a photocatalyst for selective aerobic oxidation of amines to corresponding imines.

Features and Benefits

Possesses improved photocatalytic activity.

Other Notes

Contains ~5% anatase

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiwei Zhang et al.
Scientific reports, 10(1), 5562-5562 (2020-03-30)
Autophagy and NF-κB signaling are involving in the process of Particle Disease, which was caused by the particles released from friction interface of artificial joint, implant materials of particle reinforced composite, scaffolds for tissue engineering, or material for drug delivery.
D Minetto et al.
Environment international, 66, 18-27 (2014-02-11)
The innovative properties of nanomaterials make them suitable for various applications in many fields. In particular, TiO2 nanoparticles (nTiO2) are widely used in paints, in cosmetics and in sunscreens that are products accessible to the mass market. Despite the great
Alessia D'Agata et al.
Nanotoxicology, 8(5), 549-558 (2013-05-24)
Marine bivalves (Mytilus galloprovincialis) were exposed to titanium dioxide (10 mg L(-1)) either as engineered nanoparticles (nTiO2; fresh, or aged under simulated sunlight for 7 days) or the bulk equivalent. Inductively coupled plasma-optical emission spectrometry analyses of mussel tissues showed
Susan C Tilton et al.
Nanotoxicology, 8(5), 533-548 (2013-05-11)
The growing use of engineered nanoparticles (NPs) in commercial and medical applications raises the urgent need for tools that can predict NP toxicity. Global transcriptome and proteome analyses were conducted on three human cell types, exposed to two high aspect
Nabila Haddou et al.
Chemosphere, 107, 304-310 (2014-01-28)
The Gliding Arc Discharge (GAD) is an efficient non-thermal plasma technique able to degrade organic compounds dispersed in water at atmospheric pressure. The degradation of the organometallic lead acetate (PbAc) in aqueous solution was performed by two distinct plasmageneous processes:

Questions

1–9 of 9 Questions  
  1. Do you know the shape of the particles ? And how wide is the particle distribution ?

    1 answer
    1. The shape of the particles in this product is not controlled, and the particle size distribution has not been tested. Therefore, specific information regarding the particle shape and distribution is not available.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  4. Do you know the particle size distribution of Titania powder #224227

    1 answer
    1. The specification for particle size is < 5 μm. The average particle size will be recorded on the lot specific certificate of analysis. The particle size distribution is not determined.

      See the link below to review a sample Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/224227-100G0000213989.pdf

      Helpful?

  5. What is the solubility of Product 224227, Titanium(IV) oxide, rutile?

    1 answer
    1. We do not test the solubility of this product. However, according to the chemicals encyclopedia published by the Royal Society of Chemistry, 13th Edition, this product is insoluble in water, HCl, HNO3 and dilute H2SO4. It is expected to be soluble in hot concentrated H2SO4 and hydrofluoric acid.

      Helpful?

  6. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  7. What does "Trace Metal Basis" mean?

    1 answer
    1. For this product, the purity is determined by testing total metallic impurities. Our specifications only allow for a maximum of 1000 ppm. On the Certificate of Analysis, you will be able to see which metals were tested and the amount.

      Helpful?

  8. How much anatase is present in Product 224227, Titanium(IV) oxide, rutile?

    1 answer
    1. We do not determine the amount of anatase form present. However, according to our supplier, this product may have a maximum of 10% anatase present.

      Helpful?

  9. What is the surface area of Product Product 224227, Titanium(IV) oxide, rutile?

    1 answer
    1. According to our supplier, the surface area of this product is about 2-3 m2/g.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service