Skip to Content
Merck
All Photos(2)

Key Documents

223913

Sigma-Aldrich

Ascarite®

Sodium hydroxide-coated silica, 8-20 mesh

Synonym(s):

Ascarite CO2 adsorbent

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

granular

particle size

8-20 mesh

SMILES string

[Na+].[O-H]

InChI

[1S/Na.H2O/h;1H2/q+1;/p-1]
1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

[HEMHJVSKTPXQMS-UHFFFAOYSA-M]
HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Ascarite® consists of sodium hydroxide-coated silica. It is a carbon dioxide adsorbent.[1]

Application

Ascarite® was employed as CO2 trap for the following studies:
  • Catalytic conversion of biomass-derived feedstocks into olefins and aromatics system.[2]
  • Catalytic conversion of furans in the continuous flow fixedbed reactor.[3]
  • Catalytic testing of the zeolite samples for conversion of glucose and maple wood using a semi-batch Pyroprobe reaction system.[4]
  • Isotopic analysis of methane (CH4) samples by near-infrared (NIR) continuous-wave-cavity ring-down spectrometry (CW-CRDS).[5]

Legal Information

Ascarite is a registered trademark of Arthur H. Thomas Co.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of the 13C/12C of atmospheric CH4 using near-infrared (NIR) cavity ring-down spectroscopy.
Chen Y, et al.
Analytical Chemistry, 85(23), 11250-11257 (2013)
Respiratory exchange and evaporative water loss in the flying budgerigar.
Tucker VA.
The Journal of Experimental Biology, 48(1), 67-87 (1968)
Optimizing the aromatic yield and distribution from catalytic fast pyrolysis of biomass over ZSM-5.
Foster AJ, et al
Applied Catalysis A: General, 423, 154-161 (2012)
Catalytic conversion of biomass-derived feedstocks into olefins and aromatics with ZSM-5: the hydrogen to carbon effective ratio.
Zhang H, et al.
Energy & Environmental Science, 4(6), 2297-2307 (2011)
Production of targeted aromatics by using Diels-Alder classes of reactions with furans and olefins over ZSM-5.
Cheng Y-T and Huber GW.
Green Chemistry, 14(11), 3114-3125 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service