Skip to Content
Merck
All Photos(1)

Key Documents

737828

Sigma-Aldrich

Drierite

with indicator, 10-20 mesh

Synonym(s):

Calcium sulfate, Gypsum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaSO4
CAS Number:
Molecular Weight:
136.14
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

Quality Level

form

chunks

technique(s)

gas chromatography (GC): suitable

particle size

10-20 mesh

SMILES string

[Ca++].[O-]S([O-])(=O)=O

InChI

1S/Ca.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

OSGAYBCDTDRGGQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Drierite (CaSO4) may be employed as drying agent in the following studies:
  • For the removal of water via adsorption during the gas-chromatographic estimation of methanol, ethanol and isopropanol in biological samples.
  • As scrubber for the drying of air.
  • For the drying of nitrile samples during the calorimetric estimation of their enthalpies of vaporization.
  • For the removal of water vapor during the coulometric estimation of carbon dioxide for air pollution monitoring.

Legal Information

Drierite is a trademark of W.A. Hammond Drierite Co., LTD

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

GAS CHROMATOGRAPHIC DETERMINATION OF LOWER ALCOHOLS IN BIOLOGIC SAMPLES.
H LYONS et al.
Clinical chemistry, 10, 429-432 (1964-05-01)
The ontogeny of personality in the wild guinea pig.
Guenther A, et al.
Animal behaviour, 90, 131-139 (2014)
Respirometry: anhydrous drierite equilibrates with carbon dioxide and increases washout times.
White CR, et al.
Physiological and Biochemical Zoology : Pbz, 79(5), 977-988 (2006)
Enthalpies of vaporization of organic compounds.
Howard PB and Wadso I.
Acta Chemica Scandinavica, 24, 145-149 (1970)
The analysis of carbon monoxide in urban air at the ppm level, and the normal carbon monoxide value.
L Dubois et al.
Journal of the Air Pollution Control Association, 16(3), 135-139 (1966-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service