Skip to Content
Merck
All Photos(3)

Key Documents

223816

Sigma-Aldrich

Lithium iodide hydrate

98%

Synonym(s):

Lithium iodide (LiI), hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiI · xH2O
CAS Number:
Molecular Weight:
133.85 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

composition

Degree of hydration, 1.5-3

reaction suitability

core: lithium
reagent type: catalyst

mp

73 °C (lit.)

density

3.48 g/mL at 25 °C (lit.)

SMILES string

[Li+].[I-].[H]O[H]

InChI

1S/HI.Li.H2O/h1H;;1H2/q;+1;/p-1

InChI key

WAZWGFFJLSIDMX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for ester cleavage and decarboxylation; C-C bond forming reactions; used as source of nucleophilic iodide; used as mild Lewis acid; epoxide opening; used as an additive for organometallic-mediated transformations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Srilakshmi Krishnaveni, N.; Surendra, K. et al.
Synthesis, 4, 501-501 (2004)
Bennasar, M.-L.; Jimenez, J.-M.
Tetrahedron Letters, 37, 7653-7653 (1996)
Bajwa, J. S.; Anderson, R. C.
Tetrahedron Letters, 32, 3021-3021 (1991)
David Nicolas Østedgaard-Munck et al.
Membranes, 10(11) (2020-10-30)
Investigation of electroviscous effects is of interest to technologies that exploit transport of ions through ion exchange membranes, charged capillaries, and porous media. When ions move through such media due to a hydrostatic pressure difference, they interact with the fixed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service