Skip to Content
Merck
All Photos(3)

Key Documents

223689

Sigma-Aldrich

Bis(benzonitrile)palladium(II) chloride

greener alternative

95%

Synonym(s):

Benzonitrile, palladium complex, Bis(benzonitrile)dichloropalladium(II), Bis(benzonitrile)palladium(II) chloride, Bis(phenylnitrile)dichloropalladium, Dibenzonitrilepalladium dichloride, Dichlorobis(benzonitrile)palladium, Dichlorobis(benzonitrile)palladium(II), Dichlorobis(phenyl cyanide)palladium, Palladium dichloride bis(benzonitrile), Palladium(II) chloride bis(benzonitrile) complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CN)2PdCl2
CAS Number:
Molecular Weight:
383.57
Beilstein:
3981730
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

131 °C (lit.)

greener alternative category

SMILES string

Cl[Pd]Cl.N#Cc1ccccc1.N#Cc2ccccc2

InChI

1S/2C7H5N.2ClH.Pd/c2*8-6-7-4-2-1-3-5-7;;;/h2*1-5H;2*1H;/q;;;;+2/p-2

InChI key

WXNOJTUTEXAZLD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Bis(benzonitrile)palladium(II) chloride is a coordination compound used as a catalyst in the Suzuki Cross-Coupling reaction.


We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Find details here.

Application

Bis(benzonitrile)palladium(II) chloride can be used as a catalyst:
  • For greener amine synthesis from terminal olefins by Wacker oxidation, followed by transfer hydrogenation of the resultant imine.
  • In cross-coupling reactions and α-O-glycosidation.

Formal anti-Markovnikov hydroamination of terminal olefins

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalysts of Suzuki Cross-Coupling Based on Functionalized Hyper-cross-linked Polystyrene: Influence of Precursor Nature
Nemygina N, et al.
Organic Process Research & Development, 20(8), 1453-1460 (2019)
Wei Shi et al.
Chemical communications (Cambridge, England), (23), 2342-2344 (2007-09-12)
A direct alkynylation of readily available alpha-halo esters and amides with high yields is described herein; a distinct switch from diyne formation to alkynylation products was attained under neutral conditions.
Brandon P Schuff et al.
Organic letters, 9(16), 3173-3176 (2007-07-10)
A novel method for palladium-catalyzed stereoselective formation of alpha-O-glycosides has been developed. This strategy relies on the palladium-biaryl phosphine catalyst-glycal donor complexation to control the anomeric selectivity. It does not depend on the nature of the protecting groups on the
Synthesis, 803-803 (1992)
Shih-Chieh Yeh et al.
Polymers, 11(12) (2019-12-11)
A crucial polymer intermediate, 4-[1-(4-hydroxyphenyl)cyclopentyl]-phenol (bisphenol CP), was developed from dicyclopentadiene (DCPD), a key byproduct of the C5 fraction in petrochemicals. On the basis of bisphenol CP, a diamine, 4,4'-((cyclopentane-1,1-diylbis(4,1-phenylene))bis(oxy))-dianiline (cyclopentyl diamine; CPDA) was subsequently obtained through a nucleophilic substitution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service