219827
(S)-(−)-2-Hydroxyisocaproic acid
98%
Synonym(s):
2-Hydroxy-4-methylvaleric acid, L-2-Hydroxy-4-methylpentanoic acid, L-Leucic acid
Select a Size
Select a Size
About This Item
Recommended Products
Quality Level
Assay
98%
form
powder
optical activity
[α]22/D −26.3°, c = 1 in 1 M NaOH
mp
78-80 °C (lit.)
functional group
carboxylic acid
hydroxyl
SMILES string
CC(C)C[C@H](O)C(O)=O
InChI
1S/C6H12O3/c1-4(2)3-5(7)6(8)9/h4-5,7H,3H2,1-2H3,(H,8,9)/t5-/m0/s1
InChI key
LVRFTAZAXQPQHI-YFKPBYRVSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service