Skip to Content
Merck
All Photos(2)

Key Documents

217905

Sigma-Aldrich

Copper(II) fluoride

98%

Synonym(s):

Cupric fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuF2
CAS Number:
Molecular Weight:
101.54
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

Quality Level

Assay

98%

form

powder

loss

<0.5% loss on drying, 105 °C, 1h

mp

950 °C (dec.) (lit.)

density

4.23 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

F[Cu]F

InChI

1S/Cu.2FH/h;2*1H/q+2;;/p-2

InChI key

GWFAVIIMQDUCRA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Copper fluoride is a monoclinic weak ferromagnet. The structural properties were investigated by neutron diffraction.

Application

Copper fluoride can act as an intermediate in the green synthesis of fluoro aromatics.

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T Akimoto et al.
Shika zairyo, kikai = Journal of the Japanese Society for Dental Materials and Devices, 8(3), 425-430 (1989-05-01)
As a model experiment to understand the mechanism of adhesion of the MMA/TBBO resin to dentin, polymerization of MMA/TBBO was investigated in the presence of a collagen sheet treated with the aqueous citric acid (CA) solutions of copper fluoride (CF)
T Akimoto et al.
Shika zairyo, kikai = Journal of the Japanese Society for Dental Materials and Devices, 9(2), 320-325 (1990-03-01)
As a model experiment to understand the mechanism of adhesion of the MMA-TBBO resin to dentin, MMA-TBBO was polymerized using a decalcified dentin sheet treated with aqueous citric acid (CA) solutions of copper fluoride (CF), ferric fluoride (FF), or ferric
J G de Oliveira Cordeiro
The Bulletin of Tokyo Medical and Dental University, 42(3), 105-116 (1995-12-01)
The aim of this investigation was to evaluate the effect of topical applications of various fluoride compounds on the development of root surface caries in hamsters. Male golden hamsters (n = 115) were divided into 7 groups and were given
Daisuke Tomita et al.
Chemistry, an Asian journal, 1(1-2), 161-166 (2007-04-19)
A new method for the catalytic enantioselective alkenylation and arylation of aldehydes involves the activation of alkenyl and aryl boronates by a catalytic amount of the Cu(I)F-DTBM-segphos complex through transmetalation, generating novel alkenyl and aryl copper species. These reagents act
"A greener" synthetic route for fluoroaromatics via copper (II) fluoride.
Subramanian MA and Manzer LE
Science, 297(5587), 1665-1665 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service