Skip to Content
Merck
All Photos(1)

Key Documents

19670

Sigma-Aldrich

4-tert-Butylcatechol

≥97.0% (HPLC)

Synonym(s):

4-(1,1-Dimethylethyl)benzene-1,2-diol, 4-tert-Butylcatechin, 4-tert-Butylpyrocatechol, 4-TBC, p-tert-Butylcatechol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H3-1,2-(OH)2
CAS Number:
Molecular Weight:
166.22
Beilstein:
2043335
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥97.0% (HPLC)

Quality Level

Assay

≥97.0% (HPLC)

bp

285 °C (lit.)

mp

52-55 °C (lit.)
53-58 °C

solubility

methanol: soluble 1 g/10 mL, clear, colorless to slightly yellow

SMILES string

CC(C)(C)c1ccc(O)c(O)c1

InChI

1S/C10H14O2/c1-10(2,3)7-4-5-8(11)9(12)6-7/h4-6,11-12H,1-3H3

InChI key

XESZUVZBAMCAEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-tert-Butylcatechol, in the presence of O2 catalyzed by tyrosinase, yields 4-tert-butyl-o-benzoquinone. Electrochemical oxidation of 4-tert-butylcatechol in the presence of 4-hydroxycoumarin as nucleophile has been studied using cyclic voltammetry and controlled-potential coulometry. It undergoes electrochemical trimerization via anodic oxidation and mechanism of trimerization has been studied using cyclic voltammetry and controlled-potential coulometry.

Application

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanistic study of electrochemical oxidation of 4-tert-butylcatechol: A facile electrochemical method for the synthesis of new trimer of 4-tert-butylcatechol.
Nematollahi D, et al.
Electrochimica Acta, 49(15), 2495-2502 (2004)
Electrochemical study of 3, 4-dihydroxybenzoic acid and 4-tert-butylcatechol in the presence of 4-hydroxycoumarin application to the electro-organic synthesis of coumestan derivatives.
Golabi SM and Nematollahi D.
Journal of Electroanalytical Chemistry, 430(1), 141-146 (1997)
T Kawashima et al.
The Journal of investigative dermatology, 82(1), 53-56 (1984-01-01)
4-Tertiary butyl catechol (TBC) causes depigmentation in humans and animals and stimulates formation of pheomelanosomes. In this study, we investigated the effects of noncytotoxic doses of TBC on glutathione S-transferase (GST) activity in the skin of Uscd strain mice and
María José Peñalver et al.
Biochimica et biophysica acta, 1597(1), 140-148 (2002-05-16)
The stoichiometry of oxygen consumption during tyrosinase-catalyzed oxidation of an o-diphenol (4-tert-butylcatechol, TBC) and a monophenol (4-tert-butylphenol, TBP) has been determined. At high [substrate]/[enzyme] ratios, in the case of o-diphenols, the stoichiometry of the enzyme-catalyzed reaction was always 1 O(2)/2
Sirley V Pereira et al.
The Analyst, 136(22), 4745-4751 (2011-10-11)
In this article we report the development of an integrated microfluidic system coupled to a screen-printed carbon electrode (SPCE) applied to the quantitative determination of IgG specific antibodies present in serum samples of patients that suffer from Chagas disease. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service