Skip to Content
Merck
All Photos(1)

Key Documents

196479

Sigma-Aldrich

2-Phenyl-4-quinolinecarboxylic acid

99%

Synonym(s):

2-Phenylcinchoninic acid, Cinchophen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11NO2
CAS Number:
Molecular Weight:
249.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

mp

214-215 °C (lit.)

solubility

alcohol: soluble 1g in 120ml(lit.)
chloroform: soluble 1g in 400ml(lit.)
diethyl ether: soluble 1g in 100ml(lit.)
water: insoluble(lit.)

SMILES string

OC(=O)c1cc(nc2ccccc12)-c3ccccc3

InChI

1S/C16H11NO2/c18-16(19)13-10-15(11-6-2-1-3-7-11)17-14-9-5-4-8-12(13)14/h1-10H,(H,18,19)

InChI key

YTRMTPPVNRALON-UHFFFAOYSA-N

General description

2-Phenyl-4-quinolinecarboxylic acid on reaction with 5-aryl-2,3-dihydro-2,3-furandiones yields β-aroylpyruvoyl hydrazide of 2-phenyl-4-quinolinecarboxylic acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[THERAPY OF GOUTY ARTHROPATHY].
V MECCOLI
La Clinica terapeutica, 27, 517-523 (1963-12-15)
[Atophan intoxication].
W ROSE
Archiv fur Toxikologie, 19, 158-163 (1961-01-01)
Structure-action relationship among drugs acting on connective tissues (anti-rheumatic agents).
M W WHITEHOUSE
Nature, 194, 984-985 (1962-06-09)
[DISTRIBUTION AND CONTENT OF RIBONUCLEOPROTEINS IN THE SMALL INTESTINE MUCOUS MEMBRANE IN CHRONIC EXPERIMENTAL STOMACH ULCER IN DOGS].
S A DALIMOV et al.
Meditsinskii zhurnal Uzbekistana, 39, 50-53 (1963-08-01)
[Gout].
Y OSHIMA
Naika. Internal medicine, 10, 1101-1102 (1962-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service