Skip to Content
Merck
All Photos(2)

Key Documents

460621

Sigma-Aldrich

4-Methoxypyridine

97%

Synonym(s):

γ-Methoxypyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.516 (lit.)

bp

108-111 °C/65 mmHg (lit.)

density

1.075 g/mL at 25 °C (lit.)

SMILES string

COc1ccncc1

InChI

1S/C6H7NO/c1-8-6-2-4-7-5-3-6/h2-5H,1H3

InChI key

XQABVLBGNWBWIV-UHFFFAOYSA-N

General description

4-Methoxypyridine has been prepared from 4-methoxypyridine-N-oxide, via catalytic hydrogenation. Ortho lithiation of 4-methoxypyridine using mesityllithium as the metalating base has been studied.

Application

4-Methoxypyridine was used as a starting reagent for the stereocontrolled synthesis of (±)-pumiliotoxin C and (±)-lasubine II. It was also used in an efficient construction of dihyropyridin-4-ones, which serves as potential ligands for neuronal nicotinic acetycholine receptors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tetrahedron, 60, 4861-4861 (2004)
Toxic seizures in children: case scenarios and treatment strategies.
Michael Shannon et al.
Pediatric emergency care, 19(3), 206-210 (2003-06-19)
Addition of Grignard reagents to 1-acyl-4-methoxypyridinum salts. An approach to the synthesis of quinolizidinones.
Connns DL and Brown JD.
Tetrahedron Letters, 27(38), 4549-4552 (1986)
Iu Iu Vol'dman et al.
Bioorganicheskaia khimiia, 15(8), 1044-1055 (1989-08-01)
Acceleration of substrate longitudinal relaxation (T1) was used to study cytochrome P-450-aminopyrine (1st type substrate) and P-450-4-methoxypyridine (2nd type substrate) complexes. Dissociation constant, T1 and/or residence time of substrate in the complex can be obtained from the dependence of T1
A highly stereocontrolled, four-step synthesis of (?)-lasubine II.
Brown JD, et al.
Journal of the American Chemical Society, 110(22), 7445-7447 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service