Skip to Content
Merck
All Photos(1)

Documents

134775

Sigma-Aldrich

2-Acetonaphthone

99%

Synonym(s):

2′-Acetonaphthone, 2-Acetylnaphthalene, Methyl 2-naphthyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7COCH3
CAS Number:
Molecular Weight:
170.21
Beilstein:
774965
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

300-301 °C (lit.)

mp

52-56 °C (lit.)

SMILES string

CC(=O)c1ccc2ccccc2c1

InChI

1S/C12H10O/c1-9(13)11-7-6-10-4-2-3-5-12(10)8-11/h2-8H,1H3

InChI key

XSAYZAUNJMRRIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Acetonaphthone undergoes efficient photoreduction in the presence of tri-n-butylstannane as hydrogen donor. It is solubilized in air-saturated sodium dodecyl sulphate micelles in D2O or H2O by pulsed nitrogen laser photolysis for triplet sensitized production of singlet oxygen.

Application

2-Acetonaphthone was used in direct time-resolved studies on singlet molecular oxygen phosphorescence in heterogeneous silica gel/cyclohexane systems.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

334.4 °F - DIN 51758

Flash Point(C)

168 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quenching of singlet molecular oxygen (1. DELTA. gO2) in silica gel/cyclohexane heterogeneous systems. A direct time-resolved study.
Iu KK and Thomas JK.
Journal of the American Chemical Society, 112(9), 3319-3325 (1990)
Lifetime and reactivity of singlet oxygen in an aqueous micellar system: a pulsed nitrogen laser study.
Gorman AA and Rodgers MAJ.
Chemical Physics Letters, 55(1), 52-54 (1978)
Mechanisms of Photoreactions in Solution. VI. 1 Reduction of 1-Naphthaldehyde and 2-Acetonaphthone.
Hammond GS and Leermakers PA.
Journal of the American Chemical Society, 84(2), 207-211 (1962)
Russell J Chedgy et al.
Phytochemistry, 113, 149-159 (2015-01-07)
Salicinoids are phenolic glycosides (PGs) characteristic of the Salicaceae and are known defenses against insect herbivory. Common examples are salicin, salicortin, tremuloidin, and tremulacin, which accumulate to high concentrations in the leaves and bark of willows and poplars. Although their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service