Skip to Content
Merck
All Photos(1)

Documents

190780

Sigma-Aldrich

Sodium trichloroacetate

97%

Synonym(s):

NaTCA, TCA sodium salt, Trichloroacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl3CO2Na
CAS Number:
Molecular Weight:
185.37
Beilstein:
3773249
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

SMILES string

[Na+].[O-]C(=O)C(Cl)(Cl)Cl

InChI

1S/C2HCl3O2.Na/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1

InChI key

SAQSTQBVENFSKT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium trichloroacetate with trichloroacetic acid (TCA) undergoes rapid decarboxylation in dimethylformamide (DMF) in the presence of aldehydes to form nucleophilic trichloromethyl anion, which then adds to the aldehydes to form trichloromethyl carbinols. It can also undergo thermal decarboxylation in aprotonic solvents in the presence of olefins to generate dichlorocarbene as an intermediate.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre E Galand et al.
Molecular ecology, 24(23), 5785-5795 (2015-08-21)
Understanding the link between community diversity and ecosystem function is a fundamental aspect of ecology. Systematic losses in biodiversity are widely acknowledged but the impact this may exert on ecosystem functioning remains ambiguous. There is growing evidence of a positive
Paul L A M Corstjens et al.
Parasitology, 141(14), 1841-1855 (2014-06-17)
The potential of various quantitative lateral flow (LF) based assays utilizing up-converting phosphor (UCP) reporters for the diagnosis of schistosomiasis is reviewed including recent developments. Active infections are demonstrated by screening for the presence of regurgitated worm antigens (genus specific
The thermal decarboxylation of alkali trichloroacetates in aprotic solvents. II. Decarboxylation in the presence of olefins.
Wagner WM
Rec. Trav. Chim., 80(7), 740-746 (1961)
Two effective procedures for the synthesis of trichloromethyl ketones, useful precursors of chiral ?-amino and ?-hydroxy acids.
Corey EJ
Tetrahedron Letters, 33(24), 3435-3438 (1992)
Arun A Azad et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(10), 2315-2324 (2015-02-26)
Although novel agents targeting the androgen-androgen receptor (AR) axis have altered the treatment paradigm of metastatic castration-resistant prostate cancer (mCRPC), development of therapeutic resistance is inevitable. In this study, we examined whether AR gene aberrations detectable in circulating cell-free DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service