Skip to Content
Merck
All Photos(1)

Key Documents

189960

Sigma-Aldrich

2-Methylbenzhydrol

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH(C6H5)OH
CAS Number:
Molecular Weight:
198.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98%

form

solid

bp

323 °C (lit.)

mp

93-95 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

Cc1ccccc1C(O)c2ccccc2

InChI

1S/C14H14O/c1-11-7-5-6-10-13(11)14(15)12-8-3-2-4-9-12/h2-10,14-15H,1H3

InChI key

MXHXXJOHFRHBFB-UHFFFAOYSA-N

General description

2-Methylbenzhydrol on etherification with tropine yields tropinyl 2-methyl-benzhydryl ether hydrobromide (BS 6825)[1]. It also undergoes reduction in acetic acid containing iodine and aqueous hypophosphorous acid to yield diphenylmethanes[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of alkyl substitution in drugs. IV. Pharmacological properties of tropinyl 2-methylbenzhydryl ether hydrobromide (BS 6825).
A B FUNCKE et al.
Journal of medicinal and pharmaceutical chemistry, 4, 215-224 (1961-09-08)
Further studies on the reduction of benzylic alcohols by hypophosphorous acid/iodine.
Gordon PE, et al.
ARKIVOC (Gainesville, FL, United States), 6, 393-400 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service