Skip to Content
Merck
All Photos(3)

Documents

189421

Sigma-Aldrich

Polycaprolactone diol

average Mn ~2,000

Synonym(s):

Polycaprolactone oxydiethylene ester, polycaprolactone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H8O3(C6H10O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid (waxy)

Quality Level

mol wt

average Mn ~2,000

transition temp

softening point 50 °C

density

1.071 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

Polycaprolactone diol (PCL-diol), along with polytetrahydrofuran diol (PTHF-diol) and ceric ammonium nitrate may be used to polymerize methyl methacrylate (MMA). The product may be used to prepare biocompatible poly(ester urethane)urea (PEUU) and as a precursor for polyurethane (PU) thermoplastic polymer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication of biodegradable elastomeric scaffolds with sub?micron morphologies.
Stankus J J, et al.
Journal of Biomedical Materials Research Part A, 70(4), 603-614 (2004)
Preparation, characterization and properties of polycaprolactone diol-functionalized multi-walled carbon nanotube/thermoplastic polyurethane composite.
Jing Q, et al.
Composites Part A: Applied Science and Manufacturing, 70, 8-15 (2015)
Ceric ion initiation of methyl methacrylate using polytetrahydrofuran diol and polycaprolactone diol.
Arslan H and Hazer B
European Polymer Journal, 35(8), 1451-1455 (1999)
Shih-Heng Chen et al.
International journal of molecular sciences, 20(7) (2019-04-04)
Peritendinous adhesion is a major complication after tendon injury and the subsequent repairs or reconstructions. The degree of adhesion can be reduced by the interposition of a membranous barrier between the traumatized tendon and the surrounding tissue. In the present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service