189022
2-Methyl-2-nitropropane
99%
Synonym(s):
1,1-Dimethyl-1-nitroethane, 2-Methyl-2-nitropropane, 2-Nitro-2-methylpropane, 2-Nitroisobutane, Trimethylnitromethane
About This Item
Recommended Products
Quality Level
Assay
99%
refractive index
n20/D 1.4 (lit.)
bp
126-127 °C (lit.)
density
0.95 g/mL at 25 °C (lit.)
SMILES string
CC(C)(C)[N+]([O-])=O
InChI
1S/C4H9NO2/c1-4(2,3)5(6)7/h1-3H3
InChI key
AIMREYQYBFBEGQ-UHFFFAOYSA-N
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
66.2 °F - closed cup
Flash Point(C)
19 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service