Skip to Content
Merck
All Photos(1)

Key Documents

259497

Sigma-Aldrich

1-Nitrohexane

98%

Synonym(s):

1-Nitrohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5NO2
CAS Number:
Molecular Weight:
131.17
Beilstein:
1749466
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.423 (lit.)

bp

91-92 °C/24 mmHg (lit.)

density

0.94 g/mL at 25 °C (lit.)

SMILES string

CCCCCC[N+]([O-])=O

InChI

1S/C6H13NO2/c1-2-3-4-5-6-7(8)9/h2-6H2,1H3

InChI key

FEYJIFXFOHFGCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The electroreduction of 1-nitrohexane in the absence and presence of n-octanol and n-decanol has been studied.

Application

1-Nitrohexane has been used in preparation of silyl nitronates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silyl nitronates: improved nitro-aldol reactions and reductive routes to 2-aminoalcohols.
Colvin EW and Seebach D.
Journal of the Chemical Society. Chemical Communications, 16, 689-691 (1978)
Anomalous Influence of Adsorbed Higher Aliphatic Alcohols on Electroreduction of Nitromethane and 1-Nitrohexane on Mercury Electrodes.
Gromulska A, et al.
Electroanalysis, 11(9), 595-600 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service