Skip to Content
Merck
All Photos(1)

Documents

185779

Sigma-Aldrich

2-Chlorophenol

≥99%

Synonym(s):

o-Chlorophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4OH
CAS Number:
Molecular Weight:
128.56
Beilstein:
1905114
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

form

liquid

refractive index

n20/D 1.558 (lit.)

bp

175-176 °C (lit.)

mp

8 °C (lit.)

solubility

alcohol: freely soluble
diethyl ether: freely soluble
water: slightly soluble

density

1.241 g/mL at 25 °C (lit.)

SMILES string

Oc1ccccc1Cl

InChI

1S/C6H5ClO/c7-5-3-1-2-4-6(5)8/h1-4,8H

InChI key

ISPYQTSUDJAMAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetics of 2-chlorophenol oxidation in near-critical and supercritical water in a high-pressure plug-flow reactor has been investigated. The photocatalytic degradation of 2-chlorophenol in aqueous solution has been studied using Co-doped TiO2 nanoparticles catalyst.

2-Chlorophenol is used as a reactant in Suzuki coupling and chlorination reactions and also participates in oxidation reactions to form corresponding quinones.

Application

2-Chlorophenol was used in the synthesis of polychlorinated dibenzo-p-dioxins and dibenzofurans via Cu(II)O-mediated reaction in a packed bed reactor.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photocatalytic degradation of 2-chlorophenol by Co-doped TiO2 nanoparticles.
Barakat MA, et al.
Applied Catalysis. B, Environmental, 57(1), 23-30 (2005)
Oxidation of 2-chlorophenol in water by ultrasound/Fenton method
Lin JG
Journal of Environmental Engineering (New York, N.Y.), 126(2) (2000)
The use of polymeric sulfides as catalysts for the para-regioselective chlorination of phenol and 2-chlorophenol
Smith K, et al.
Journal of Sulfur Chemistry, 41(1), 1-12 (2020)
2-Chlorophenol oxidation in supercritical water: Global kinetics and reaction products.
Li R, et al.
AIChE Journal, 39(1), 178-187 (1993)
Bulky 2, 6-dibenzhydryl-4-methylphenyl ?-diiminato derived complexes of Pd (II) and Cu (II): Efficient catalysts for Suzuki coupling and alcohol oxidation
Saxena P, et al.
Journal of Organometallic Chemistry, 868, 76-85 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service