Skip to Content
Merck
All Photos(2)

Key Documents

182125

Sigma-Aldrich

Poly(hexyl methacrylate) solution

average Mw ~400,000 by GPC, in toluene

Synonym(s):

Hexyl methacrylate homopolymer, Poly(1-hexyl methacrylate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H18O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

1.2 (37 °C, vs air)

form

liquid

mol wt

average Mw ~400,000 by GPC

concentration

in toluene

refractive index

n20/D 1.481

transition temp

Tg −5 °C

density

1.007 g/mL at 25 °C

SMILES string

CCCCCCOC(=O)C(C)=C

InChI key

LNCPIMCVTKXXOY-UHFFFAOYSA-N

General description

Poly(hexadecyl methacrylate) belongs to a class of long chain of poly (alkyl) methacrylate. It can be dissolved in benzene and heptane.

Application

Poly(hexyl methacrylate)(PHMA) may be used to prepare amphiphilic block copolymers with -poly(ethylene oxide) (PEO). These block copolymers may be used as structure directing agents for the preparation organic-inorganic hybrid materials from organic, volatile solvents.

Other Notes

Unit weight does not include weight of solvent.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanostructure and Shape Control in Polymer?Ceramic Hybrids from Poly (ethylene oxide)?block?Poly (hexyl methacrylate) and Aluminosilicates Derived from Them.
Renker S, et al.
Macromolecular Chemistry and Physics, 205(8), 1021-1030 (2004)
Synthesis and Characterization of Amphiphilic Poly (ethylene oxide)?block?poly (hexyl methacrylate) Copolymers.
Mahajan S, et al.
Macromolecular Chemistry and Physics, 204(8), 1047-1055 (2003)
Polymers: a property database. (2008)
Physical properties of polymers handbook (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service