All Photos(2)
About This Item
Linear Formula:
HC≡CC6H10NH2
CAS Number:
Molecular Weight:
123.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.482 (lit.)
bp
65-66 °C/20 mmHg (lit.)
density
0.913 g/mL at 25 °C (lit.)
SMILES string
NC1(CCCCC1)C#C
InChI
1S/C8H13N/c1-2-8(9)6-4-3-5-7-8/h1H,3-7,9H2
InChI key
GDKOYYDQISQOMH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
1-Ethynylcyclohexylamine was used in the synthesis of bicyclic and tetracyclic compounds. It was also used as a substrate in the reaction of a series of alkynes with mercury(II)acetate.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
107.6 °F - closed cup
Flash Point(C)
42 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bassetti M and Floris B.
J. Chem. Soc. Perkin Trans. II, 2, 227-233 (1988)
Cyclization reaction of cyclopentadienone with prop-2-yn-1-ol in priority to Diels?Alder reaction.
Tetrahedron Letters, 47(25), 4235-4239 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service