Skip to Content
Merck
All Photos(1)

Key Documents

150223

Sigma-Aldrich

N-Methylpropargylamine

95%

Synonym(s):

3-Methylamino-1-propyne

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCH2NHCH3
CAS Number:
Molecular Weight:
69.11
Beilstein:
1071242
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

impurities

2-3% water

refractive index

n20/D 1.432 (lit.)

bp

82-84 °C (lit.)

density

0.819 g/mL at 25 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CNCC#C

InChI

1S/C4H7N/c1-3-4-5-2/h1,5H,4H2,2H3

InChI key

HQFYIDOMCULPIW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

N-Methylpropargylamine was used in the preparation of dimethyl 4-benzylidene-1-methyl-2-phenyl-pyrrolidine-3,3-dicarboxylate.
Reacts with supercritical carbon dioxide to give 5-alkyidene oxazolidinones.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Three-component one-pot synthesis of functionalised Z-4-benzylidene (and alkenylidene) pyrrolidines.
Azoulay S, et al.
Tetrahedron Letters, 43(51), 9311-9314 (2002)
D Ratnaweera et al.
Allergy, 61(8), 1019-1020 (2006-07-27)
A community-based study in the London Borough of Hounslow, which included patients in our District General Hospital (DGH) Paediatric Department, found that most families who had been prescribed adrenaline auto-injectors could not use them properly. This prompted the establishment of
Fan Yang et al.
Nature chemical biology, 14(9), 876-886 (2018-08-19)
Signals from 800 G-protein-coupled receptors (GPCRs) to many SH3 domain-containing proteins (SH3-CPs) regulate important physiological functions. These GPCRs may share a common pathway by signaling to SH3-CPs via agonist-dependent arrestin recruitment rather than through direct interactions. In the present study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service