Skip to Content
Merck
All Photos(3)

Key Documents

164747

Sigma-Aldrich

Europium tris[3-(heptafluoropropylhydroxymethylene)-(+)-camphorate]

98%

Synonym(s):

Eu(hfc)3, Europium(III) tris[3-(heptafluoropropylhydroxymethylene)-d-camphorate], Tris[3-(heptafluoropropylhydroxymethylene)-d-camphorato]europium(III)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H42EuF21O6
CAS Number:
Molecular Weight:
1193.71
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

optical activity

[α]20/D +158.0°, c = 1 in chloroform

technique(s)

NMR: suitable

mp

156-158 °C (lit.)

functional group

fluoro
ketone

SMILES string

CC1(C)C2CC[C@@]1(C)C(=O)\C2=C(\O[Eu](O\C(=C3\C4CC[C@@](C)(C3=O)C4(C)C)C(F)(F)C(F)(F)C(F)(F)F)O\C(=C5/C6CC[C@@](C)(C5=O)C6(C)C)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/3C14H15F7O2.Eu/c3*1-10(2)6-4-5-11(10,3)8(22)7(6)9(23)12(15,16)13(17,18)14(19,20)21;/h3*6,23H,4-5H2,1-3H3;/q;;;+3/p-3/b3*9-7+;/t3*6-,11+;/m111./s1

InChI key

VGLKHVQPWGFXEG-PSMAIOHISA-K

Application

Eu(hfc)3 may be used as an optically active NMR shift reagent in the 1H NMR analysis to determine the following:
  • Enantiomeric purities of the precursors of trisporic acids A, B and related products.
  • Optical purity of racemic 6,7-dichloro-5-methoxy-2-methyl-2-phenyl-1 indanone.
  • Epimeric purity of 4-methylsulfinylbutylglucosinolate.
For use as a Lanthanide shift reagent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective synthesis of the white key intermediate for the synthesis of trisporic acids.
Bacigaluppo JA, et al.
Tetrahedron Asymmetry, 5(10), 1877-1880 (1994)
Determination of the absolute configuration of the glucosinolate methyl sulfoxide group reveals a stereospecific biosynthesis of the side chain.
Vergara F, et al.
Phytochemistry, 69(15), 2737-2742 (2008)
Optical purity determination and 1H-NMR spectral simplification with lanthanide shift reagents-VIII. An indacrinone precursor, 6, 7-dichloro-5-methoxy-2-methyl-2-phenyl-1-indanone.
Hatzis A and Rothchild R.
Journal of Pharmaceutical and Biomedical Analysis, 4(4), 443-449 (1986)
Pirkle et al.
The Journal of Organic Chemistry, 44, 1025-1025 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service