Skip to Content
Merck
All Photos(2)

Key Documents

161950

Sigma-Aldrich

1,3-Diethyl-2-thiobarbituric acid

99%

Synonym(s):

1,3-Diethyl-2-sulfanylidene-1,3-diazinane-4,6-dione, 1,3-Diethyldihydro-2-thioxo-4,6(1H,5H)-pyrimidinedione, 1,3-Diethylthiobarbituric acid, N,N′-Diethyl-2-thiobarbituric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O2S
CAS Number:
Molecular Weight:
200.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

mp

109-112 °C (lit.)

solubility

1 M NaOH: soluble 50 mg/mL, clear, colorless to light yellow

SMILES string

CCN1C(=O)CC(=O)N(CC)C1=S

InChI

1S/C8H12N2O2S/c1-3-9-6(11)5-7(12)10(4-2)8(9)13/h3-5H2,1-2H3

InChI key

SHBTUGJAKBRBBJ-UHFFFAOYSA-N

Application

1,3-Diethyl-2-thiobarbituric acid (DETBA) was used as coinitiator during the photopolymerization of dental materials. It was used as reagent in electrochemical oxidation of 3,4-dihydroxybenzoic acid to yield benzofuro[2,3-d]pyrimidine derivatives. It was used in DETBA assay for determination of nicotine metabolites in human urine by HPLC.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jadwiga Turło et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48(4), 1085-1091 (2010-02-09)
Preparations derived from Lentinula edodes (Berk.) Pegl. mycelium are worldwide used as dietary supplements containing compounds active as immune system enhancers, demonstrating chemopreventive and anticancer activity. L. edodes mycelium enriched with organic forms of selenium like selenized yeast possess putative
R F Smith et al.
Clinical chemistry, 44(2), 275-280 (1998-02-25)
The performance of a simple colorimetric assay for urinary nicotine metabolites to assess smoking status in diabetic subjects (n = 251) was investigated. Several variations of the colorimetric assay and a qualitative extraction procedure were evaluated in comparison with a
T Sakai et al.
Journal of chromatography. B, Biomedical sciences and applications, 726(1-2), 313-316 (1999-05-29)
The reaction conditions of 1,3-diethyl-2-thiobarbituric acid (DETBA)-malonaldehyde (MA) adduct formation were examined in order to analyze MA in fish tissue by high-performance liquid chromatography. A reaction mixture containing 4 mM butyl hydroxytoluene was heated at 100 degrees C for 150
H Peach et al.
IARC scientific publications, (74)(74), 183-193 (1986-01-01)
Three novel colorimetric methods of detecting urinary nicotine metabolites called the barbituric acid, diethylthiobarbituric acid (DETB) and DETB-extraction methods were evaluated for use as a simple cheap objective test of smoking. Urine samples were collected from 103 male smokers and
K Nakashima et al.
Biomedical chromatography : BMC, 6(2), 55-58 (1992-03-01)
The peroxyoxalate chemiluminescence(CL) detection method for the evaluation of the CL intensity of malondialdehyde(MDA) condensates with seven 2-thiobarbituric acid derivatives is described. The method consists of a flow injection technique together with a CL detection system using bis(2,4,6-trichlorophenyl) oxalate(TCPO) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service