Skip to Content
Merck
All Photos(1)

Key Documents

44977

Sigma-Aldrich

DISBAC2(3), Voltage Sensitive Probe

≥98% (HPLC)

Synonym(s):

5-[3-(1,3-Diethylhexahydro-4,6-dioxo-2-thioxo-5-pyrimidinyl)-2-propen-1-ylidene]-1,3-diethyldihydro-2-thioxo-4,6(1H,5H)-pyrimidinedione, Bis(1,3-diethylthiobarbituric acid)trimethine oxonol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N4O4S2
CAS Number:
Molecular Weight:
436.55
Beilstein:
1232950
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98% (HPLC)

fluorescence

λex 535 nm; λem 560 nm in DMSO

suitability

complies for Infrared spectrum

storage temp.

−20°C

SMILES string

CCN1C(=O)C(\C=C\C=C2\C(=O)N(CC)C(=S)N(CC)C2=O)C(=O)N(CC)C1=S

InChI

1S/C19H24N4O4S2/c1-5-20-14(24)12(15(25)21(6-2)18(20)28)10-9-11-13-16(26)22(7-3)19(29)23(8-4)17(13)27/h9-12H,5-8H2,1-4H3/b10-9+

InChI key

VJYNRXFXHKIGLT-MDZDMXLPSA-N

Application

DISBAC2(3), Voltage Sensitive Probe is used for single-cell fluorescence imaging of transmembrane conductance and potential change.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In situ study of the membrane potential in microvascular endothelial cells using a fluorescent probe.
K Miao et al.
Microvascular research, 48(1), 135-142 (1994-07-01)
M K Tanner et al.
Cytometry, 14(1), 59-69 (1993-01-01)
Immunosuppression by the fungal metabolite cyclosporin A (CsA) is characterized by functional inhibition, rather than destruction of cells. Because activation of immune cells involves intracellular signalling events associated with modulations of cell transmembrane potential (TMP), we tested the ability of
S Bouhdid et al.
Journal of applied microbiology, 109(4), 1139-1149 (2010-05-12)
To study cellular damage induced by Cinnamomum verum essential oil in Pseudomonas aeruginosa ATCC 27853 and Staphylococcus aureus ATCC 29213. The effect of cinnamon bark essential oil on these two strains was evaluated by plate counts, potassium leakage, flow cytometry
V Dall'Asta et al.
Experimental cell research, 231(2), 260-268 (1997-03-15)
Confocal laser scanning microscopy (CLSM) was employed to visualize and measure membrane potential changes in several types of cultured adherent cells, such as human fibroblasts, mouse mammary tumor C127 cells, and human saphenous vein endothelial cells, preloaded with the anionic
Mikiyo Yamaguchi et al.
Anesthesia and analgesia, 100(2), 419-426 (2005-01-28)
The membrane potential of endothelial cells is an important determinant of endothelial functions, including regulation of vascular tone. We investigated whether adenosine triphosphate-sensitive potassium (K(ATP)) channels were involved in the response of membrane potential to hyperosmolality in cultured human aorta

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service