Skip to Content
Merck
All Photos(1)

Key Documents

198234

Sigma-Aldrich

Tetrahydro-4-pyranol

98%

Synonym(s):

4-Hydroxytetrahydropyran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O2
CAS Number:
Molecular Weight:
102.13
Beilstein:
102736
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

87 °C/15 mmHg (lit.)

density

1.071 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl

SMILES string

OC1CCOCC1

InChI

1S/C5H10O2/c6-5-1-3-7-4-2-5/h5-6H,1-4H2

InChI key

LMYJGUNNJIDROI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetrahydro-4-pyranol (4-Hydroxytetrahydropyran) was used in the quantitative analysis of 3-hydroxytetrahydropyran (3-HTHP)[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancement of cyclic ether formation from polyalcohol compounds in high temperature liquid water by high pressure carbon dioxide.
Yamaguchi A, et al.
Green Chemistry, 11(1), 48-52 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service