Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.4435 (lit.)
bp
81-83 °C/15 mmHg (lit.)
density
0.884 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CCN(CC)CCCO
InChI
1S/C7H17NO/c1-3-8(4-2)6-5-7-9/h9H,3-7H2,1-2H3
InChI key
WKCYFSZDBICRKL-UHFFFAOYSA-N
Application
3-Diethylamino-1-propanol was used to develop a model system that polymerizes racemic lactide (D,L-LA).
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
150.8 °F - closed cup
Flash Point(C)
66 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of the American Chemical Society, 132(33), 11649-11657 (2010-08-03)
We report the results of a comprehensive investigation of the recently discovered stereoselective and controlled polymerization of racemic lactide (D,L-LA) using an initiator prepared in situ from indium(III) chloride (InCl(3)), benzyl alcohol (BnOH), and triethylamine (NEt(3)). Linear relationships between number-average
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service