Skip to Content
Merck
All Photos(1)

Key Documents

148601

Sigma-Aldrich

N-Methyl-p-toluenesulfonamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NHCH3
CAS Number:
Molecular Weight:
185.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

76-79 °C (lit.)

functional group

amine

SMILES string

CNS(=O)(=O)c1ccc(C)cc1

InChI

1S/C8H11NO2S/c1-7-3-5-8(6-4-7)12(10,11)9-2/h3-6,9H,1-2H3

InChI key

GWLOGZRVYXAHRE-UHFFFAOYSA-N

Application

N-Methyl-p-toluenesulfonamide was used in the synthesis of vicinal haloamino ketone derivative.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Sun et al.
Chemical biology & drug design, 75(3), 269-276 (2010-03-25)
The combinations of N-methyl-p-toluenesulfonamide/NBS and N-ethyl-p-toluenesulfonamide/NBS were found to be good nitrogen/halogen resources for the aminohalogenation of alpha,beta-unsaturated ketones in the presence of Ni(OAc)(2) as the catalyst for the synthesis of vicinal haloamino ketone derivatives. The introduction of N-alkyl groups

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service