Skip to Content
Merck
All Photos(3)

Documents

135895

Sigma-Aldrich

Dimethylthiocarbamoyl chloride

97%

Synonym(s):

1-Chloro-N,N-dimethylthioformamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCSCl
CAS Number:
Molecular Weight:
123.60
Beilstein:
635823
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

bp

90-95 °C/0.5 mmHg (lit.)

mp

39-43 °C (lit.)

solubility

chloroform: soluble 100 mg/mL, clear to very slightly hazy, very faintly yellow

storage temp.

2-8°C

SMILES string

CN(C)C(Cl)=S

InChI

1S/C3H6ClNS/c1-5(2)3(4)6/h1-2H3

InChI key

PHWISQNXPLXQRU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dimethylthiocarbamoyl chloride was used in chemoselective deoxygenation of pyridine-N-oxides. It was used in the synthesis of (±)-thia-calanolide A. It was used as starting reagent in the synthesis of dimethylthiocarbamates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A synthesis of (?)-thia-calanolide A, its resolution and in vitro biological evaluation.
Chopade AU, et al. et al.
Arabian Journal of Chemistry (2012)
Harley D Betts et al.
Molecules (Basel, Switzerland), 25(19) (2020-10-07)
Silver(I)-based coordination polymers or metal-organic frameworks (MOFs) display useful antibacterial properties, whereby distinct materials with different bonding can afford control over the release of silver(I) ions. Such silver(I) materials are comprised of discrete secondary building units (SBUs), and typically formed
Journal of Heterocyclic Chemistry, 44, 487-487 (2007)
D K Barma et al.
Organic letters, 5(25), 4755-4757 (2003-12-05)
Dimethylthiocarbamates (DMTCs), prepared from the corresponding alcohols using commercial dimethylthiocarbamoyl chloride, are spectrally simple, achiral, and nonpolar. DMTCs are moderately to highly stable to a wide range of reagents and conditions including metal hydrides, hydroboration, ylides, NaOH, HCl, organolithiums, Grignards

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service