Skip to Content
Merck
All Photos(3)

Key Documents

132306

Sigma-Aldrich

Zinc acetylacetonate hydrate

Synonym(s):

Bis(2,4-pentanedionato)zinc, Zn(acac)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn(C5H7O2)2 · xH2O
CAS Number:
Molecular Weight:
263.61 (anhydrous basis)
Beilstein:
4166200
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Zn, 23-26% EDTA titration

reaction suitability

reagent type: catalyst

mp

135-138 °C (lit.)

SMILES string

[H]O[H].CC(=O)\C=C(\C)O[Zn]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.H2O.Zn/c2*1-4(6)3-5(2)7;;/h2*3,6H,1-2H3;1H2;/q;;;+2/p-2/b2*4-3-;;

InChI key

KUJHAYOLESEVSA-SUKNRPLKSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Zinc acetylacetonate hydrate (Zn(acac)2) is a hydrated metal organic complex that is hygroscopic in nature. It is majorly used as a precursor for the synthesis of zinc based nano and microparticles.

Application

Zn(acac)2 can be in the fabrication of zinc based films and coatings in chemical vapor deposition for optoelectronic applications.

Features and Benefits

Novel MOCVD precursor for the preparation of high-crystallinity (Zn,Fe)Fe2O4 films and magnetic property measurements of these films.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zinc acetylacetonate hydrate adducted with nitrogen donor ligands: Synthesis, spectroscopic characterization, and thermal analysis
Brahma S and Shivashankar SA
Journal of Molecular Structure, 1101, 41-49 (2015)
Efficiency enhancement of organic and thin-film silicon solar cells with photochemical upconversion
Schulze TF, et al.
The Journal of Physical Chemistry C, 116(43), 22794-22801 (2012)
The synthesis of zinc oxide nanoparticles from zinc acetylacetonate hydrate and 1-butanol or isobutanol
Ambrozic G, et al.
Journal of Colloid and Interface Science, 346(2), 317-323 (2010)
A facile route to inverted polymer solar cells using a precursor based zinc oxide electron transport layer
De Bruyn P, et al.
Organic Electronics, 11(8), 1419-1422 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service