Skip to Content
Merck
All Photos(2)

Key Documents

126691

Sigma-Aldrich

Diphenic acid

97%

Synonym(s):

Biphenyl-2,2′-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO2C)C6H4C6H4-2-(CO2H)
CAS Number:
Molecular Weight:
242.23
Beilstein:
2053625
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

mp

227-229 °C (lit.)

SMILES string

OC(=O)c1ccccc1-c2ccccc2C(O)=O

InChI

1S/C14H10O4/c15-13(16)11-7-3-1-5-9(11)10-6-2-4-8-12(10)14(17)18/h1-8H,(H,15,16)(H,17,18)

InChI key

GWZCCUDJHOGOSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F H Clarke et al.
Journal of pharmaceutical sciences, 76(8), 611-620 (1987-08-01)
A convenient procedure has been developed for the determination of partition and distribution coefficients. The method involves the potentiometric titration of the compound, first in water and then in a rapidly stirred mixture of water and octanol. An automatic titrator
Pierre Thuéry et al.
Dalton transactions (Cambridge, England : 2003), 49(3), 817-828 (2019-12-20)
Diphenic acid (H2dip) has been used to synthesize nine homo- or heterometallic uranyl ion complexes under solvo-hydrothermal conditions. The diphenate ligand dip2- adopts different coordination modes, mixtures of κ2-O,O'-chelation by individual carboxylate groups, chelation involving both carboxylate groups, and bridging
Masashi Kunitake et al.
Langmuir : the ACS journal of surfaces and colloids, 21(20), 9206-9210 (2005-09-21)
Formation of adlayers of the optically active compound 1,1'-binaphthyl-2, 2'-dicarboxylic acid (BINAC) on iodine-modified Au (111) surfaces in perchloric acid was investigated by in situ scanning tunneling microscopy (STM). Highly ordered arrays formed on the surfaces via simple spontaneous adsorption
K E Hammel et al.
Applied and environmental microbiology, 58(6), 1832-1838 (1992-06-01)
The ligninolytic fungus Phanerochaete chrysosporium oxidized phenanthrene and phenanthrene-9,10-quinone (PQ) at their C-9 and C-10 positions to give a ring-fission product, 2,2'-diphenic acid (DPA), which was identified in chromatographic and isotope dilution experiments. DPA formation from phenanthrene was somewhat greater
Xiang-Wen Kong et al.
Chemistry & biodiversity, 5(9), 1743-1752 (2008-09-26)
In search of novel anticancer agents, two series of dimethyl [1,1'-biphenyl]-2,2'-dicarboxylate derivatives, 8a-8k and 9a-9k, containing both methylenedioxy and 1,3,4-thiadiazole moieties were designed and synthesized. Cytotoxicity of these compounds was evaluated in vitro against five human tumor cell lines, i.e.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service