Skip to Content
Merck
All Photos(1)

Documents

398462

Sigma-Aldrich

Gold(III) hydroxide

Synonym(s):

Gold Trihydroxide, Gold trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Au(OH)3
CAS Number:
Molecular Weight:
247.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

79-81.4% Au basis (after reduction with hydrazine dihydrate, gravimetric)

form

powder

SMILES string

O[Au](O)O

InChI

1S/Au.3H2O/h;3*1H2/q+3;;;/p-3

InChI key

WDZVNNYQBQRJRX-UHFFFAOYSA-K

Application

Synthesis of gold hydroxide complexes with N-heterocyclic carbene has been reported. A variety of perfluoroaryls, N-heterocyclic and alkynyl compounds may be synthesized by reacting gold(III) hydroxide with C–H and N–H compounds and arylboronic acids. Potassium aurate may be prepared by heating auric hydroxide with conc. KOH. 3

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclometallated gold(III) hydroxides as versatile synthons for Au?N, Au?C complexes and luminescent compounds.
Rosca DA, et al.
Chemical Communications (Cambridge, England), 48, 7247-7249 (2012)
Synthesis of gold hydroxide complex bearing N-heterocyclic carbene was reported.
Gaillard S, et al.
Chemical Communications (Cambridge, England), 46, 2742-2744 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service