Skip to Content
Merck
All Photos(1)

Key Documents

124192

Sigma-Aldrich

1,4-Butanediol diglycidyl ether

technical grade, 60%

Synonym(s):

1,4-Bis(2,3-epoxypropoxy)butane, 1,4-Bis(glycidyloxy)butane, 1,4-Bis(oxiran-2-ylmethoxy)butane, 1,4-Butylene glycol diglycidyl ether, Tetramethylene glycol diglycidyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O4
CAS Number:
Molecular Weight:
202.25
Beilstein:
115238
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

vapor pressure

~10 mmHg ( 20 °C)

form

liquid

concentration

60%

refractive index

n20/D 1.453 (lit.)

bp

266 °C (lit.)

density

1.1 g/mL at 25 °C (lit.)

SMILES string

C(CCOCC1CO1)COCC2CO2

InChI

1S/C10H18O4/c1(3-11-5-9-7-13-9)2-4-12-6-10-8-14-10/h9-10H,1-8H2

InChI key

SHKUUQIDMUMQQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Butanediol diglycidyl ether(BDDE) is a homobifunctional monomer with two epoxide groups, widely used to modify the viscosity of epoxy resins. It is also used as a cross-linking agent to synthesize polymer networks.

Application

1,4-Butanediol diglycidyl ether can be used:
  • As a cross-linking agent to prepare hyaluronic acid dermal fillers This crosslinking process enhances the gel-like consistency of the filler, making it more durable and longer-lasting.
  • As a monomer to prepare epoxy-based graphene nanocomposites that have potential applications in the field of flexible electronics, corrosion resistance coatings, and conductive adhesives. BDDE is chosen for its desirable properties such as low viscosity, good reactivity, and compatibility with graphene.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative physicochemical analysis among 1, 4-butanediol diglycidyl ether cross-linked hyaluronic acid dermal fillers
Nicola Zerbinati, et al.
Gels, 7, 139-139 (2021)
Petr Kubán et al.
Analytical chemistry, 79(14), 5462-5467 (2007-06-21)
The preparation and performance of a multilayered stationary phase for open tubular anion exchange chromatography in relatively large bore (75 microm diameter) columns are described. The inner surface of a fused-silica capillary tube is coated with up to 25 successive
D S Pepper
Molecular biotechnology, 2(2), 157-178 (1994-10-01)
Three different coupling chemistries that have been tried and tested for use in affinity chromatography are described. These methods are particularly recommended for use by workers who do not have access to, or do not wish to use, complex organic
Anabela Alves et al.
International journal of pharmaceutics, 426(1-2), 76-81 (2012-01-28)
The polysaccharide ulvan, composed of sulphated rhamnose, glucoronic and iduronic acids was used to produce polymeric membranes by solvent casting. As ulvan is soluble in water, a cross-linking step was necessary to render the membrane insoluble in water and stable
A Nicoletti et al.
Journal of materials science. Materials in medicine, 24(1), 17-35 (2012-10-12)
Serious cartilage lesions (Outerbridge III, IV) may be successfully treated with a three-layered gradient scaffold made by magnesium-doped hydroxyapatite and type I collagen, manufactured through a bio-inspired process and stabilised by a reactive bis-epoxy (1,4-butanediol diglycidyl ether, BDDGE). Each layer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service