116637
4-(Benzyloxy)aniline hydrochloride
98%
Synonym(s):
4-Aminophenyl benzyl ether hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5CH2OC6H4NH2 · HCl
CAS Number:
Molecular Weight:
235.71
Beilstein:
3633307
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
solid
mp
228 °C (dec.) (lit.)
functional group
phenyl
SMILES string
Cl.Nc1ccc(OCc2ccccc2)cc1
InChI
1S/C13H13NO.ClH/c14-12-6-8-13(9-7-12)15-10-11-4-2-1-3-5-11;/h1-9H,10,14H2;1H
InChI key
KQBDLOVXZHOAJI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
4-(benzyloxy)aniline hydrochloride can be used in chemical synthesis studies.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service