116327
2-Chloro-4-methylpyridine
98%
Synonym(s):
2-Chloro-4-picoline
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H6ClN
CAS Number:
Molecular Weight:
127.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
refractive index
n20/D 1.529 (lit.)
bp
194-195 °C (lit.)
density
1.142 g/mL at 25 °C (lit.)
functional group
chloro
SMILES string
Cc1ccnc(Cl)c1
InChI
1S/C6H6ClN/c1-5-2-3-8-6(7)4-5/h2-4H,1H3
InChI key
MZVSTDHRRYQFGI-UHFFFAOYSA-N
General description
2-Chloro-4-methylpyridine (2-Chloro-4-picoline) reacts with 3,5-bis(trifluoromethyl)phenylboronic acid to form 2-(3,5-bis(trifluoromethyl)phenyl)-4-methylpyridine by palladium-catalyzed cross-coupling reaction.
2-Chloro-4-picoline (2-Chloro-4-methylpyridine) has been used in the determination of gas-phase basicity of several relatively small organic molecules that are used in the MALDI.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Barylyuk K, et al.
Royal Society of Chemistry Advances, 2(5), 1962-1969 (2012)
Ionkin AS, et al.
Journal of Organometallic Chemistry, 692(22), 4809-4827 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service