Skip to Content
Merck
All Photos(2)

Key Documents

123080

Sigma-Aldrich

2-Amino-4-methylpyridine

99%

Synonym(s):

2-Amino-4-picoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
Beilstein:
107066
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

bp

230 °C (lit.)

mp

96-99 °C (lit.)

solubility

DMF: freely soluble
H2O: freely soluble
aliphatic hydrocarbons: slightly soluble
coal tar bases: freely soluble
lower alcohols: freely soluble
petroleum ether: slightly soluble

SMILES string

Cc1ccnc(N)c1

InChI

1S/C6H8N2/c1-5-2-3-8-6(7)4-5/h2-4H,1H3,(H2,7,8)

InChI key

ORLGLBZRQYOWNA-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

2-Amino-4-methylpyridine acts as ligand and forms methoxo-bridged copper(II) complexes.

Application

2-Amino-4-methylpyridine has been used in the synthesis of 2-amino-4-methyl­pyridinium 2-hy­droxy­benzoate.

Biochem/physiol Actions

2-Amino-4-methylpyridine inhibits the activity of inducible NO synthase isolated from mouse RAW 264.7 cells in vitro.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analgesic activity of 2-amino-4-methylpyridine, a novel NO synthase inhibitor.
E R Pettipher et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 46 Suppl 2, S135-S136 (1997-08-01)
Ravil R Petrov et al.
Bioorganic & medicinal chemistry letters, 16(18), 4946-4950 (2006-07-11)
An enkephalin analogue coupled to 'aminofentanyl' has been synthesized and tested for biological activities at the mu and delta opioid receptors. Aminofentanyl which represents a structural derivative of fentanyl has been synthesized by acylation of 1-(2-phenethyl)-4-(N-anilino)piperidine with phthaloyl protected beta-alaninyl
Eun-Ae Jo et al.
Chemical communications (Cambridge, England), (44)(44), 5779-5781 (2008-11-15)
The hydroacylation of methanol with alkenes was developed using a catalytic system consisting of Rh(I), 2-amino-4-picoline and benzoic acid; the reaction is speculated to occur by the initial N-methylation of 2-amino-4-picoline with methanol, and the subsequent dehydrogenation of the resulting
Somnath Ray Choudhury et al.
Dalton transactions (Cambridge, England : 2003), (37)(37), 7617-7624 (2009-09-18)
The crystal lattice of the three isostructural compounds , (C(6)H(8)N(2)H)(2)[M(C(3)H(2)O(4))(2)(H(2)O)(2)].4H(2)O (C(6)H(8)N(2)H = protonated 2-amino-4-picoline, M = Ni/Co/Mn, C(3)H(4)O(4) = malonate dianion; hereafter, malonate) is formed by supramolecular 2D layers. Hydrogen-bonding, pi...pi and lone pair...pi interactions play crucial role in organizing
N Kiloh et al.
European journal of pharmacology, 70(1), 53-57 (1981-03-05)
4-Methyl-2-aminopyridine (4M2AP) increased responses of isolated rat diaphragm and chick biventer cervicis nerve-muscle preparations to nerve stimulation but depressed responses to direct stimulation. Responses to acetylcholine were also increased while responses to carbachol were depressed. When tested after inhibition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service